Skip to Content
Merck
All Photos(1)

Key Documents

8.18770

Sigma-Aldrich

Cyclopentane

for synthesis

Synonym(s):

Cyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10
CAS Number:
Molecular Weight:
70.13
MDL number:
UNSPSC Code:
12352001
EC Index Number:
206-016-6
NACRES:
NA.22
Assay:
≥95% (GC)
bp:
49 °C
vapor pressure:
346 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

346 hPa ( 20 °C)

Quality Level

Assay

≥95% (GC)

form

liquid

autoignition temp.

380 °C

potency

11400 mg/kg LD50, oral (Rat)

expl. lim.

1.5-8.7 % (v/v)

bp

49 °C

mp

-93.3 °C

transition temp

flash point -42 °C

solubility

insoluble 0.156 g/L

density

0.75 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1CCCC1

InChI

1S/C5H10/c1-2-4-5-3-1/h1-5H2

InChI key

RGSFGYAAUTVSQA-UHFFFAOYSA-N

Application

  • Di-Isatropolone C, a Spontaneous Isatropolone C Dimer Derivative with Autophagy Activity: Reports on a novel cyclopentane ring formation in the study of autophagy, highlighting synthetic potential in pharmacology (Fu et al., 2024).
  • Dearomative (3+ 2) Cycloaddition of Indoles for the Stereoselective Assembly of Fully Functionalized Cyclopentanoids: Showcases the use of cyclopentane in creating complex cyclopentanoids for advanced organic synthesis (Qatran Al-Khdhairawi et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.740 - 0.745
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service