Skip to Content
Merck

Skip To

8.14767

n-Tetradecane

for synthesis

Synonym(s):

n-Tetradecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H30
CAS Number:
Molecular Weight:
198.39
UNSPSC Code:
12352001
EC Index Number:
211-096-0
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1.55 Pa ( 25 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

202 °C

potency

>5000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.5-6.5 % (v/v)

kinematic viscosity

3 cSt(40 °C)

mp

6 °C

transition temp

flash point 102 °C

solubility

<0.0001 g/L

density

0.76 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCC)CCCCCC

InChI

1S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3

InChI key

BGHCVCJVXZWKCC-UHFFFAOYSA-N

Application

  • Scaling On Transport Properties of Nanofluids for Heat Transfer Applications: Discusses the use of n-tetradecane in nanofluids for heat transfer applications, validated by experimental data (Prado et al., 2024).
  • Improvement in the freeze-thaw resistance performance of concretes: Studies the encapsulation of n-tetradecane in a composite phase change material for enhancing the freeze-thaw resistance of concrete (Li et al., 2024).
  • Density and speed of sound of liquid mixtures: Provides measurements of density and sound speed in mixtures including n-tetradecane, with implications for thermodynamic modeling (Hevia et al., 2024).
  • Thermal volume expansion: Examines thermal volume changes in n-tetradecane using multiple measurement techniques, enhancing understanding of material properties under temperature changes (Klingler et al., 2024).
  • High-Pressure Phase Behavior of Binary Systems: Investigates the phase behavior of n-tetradecane mixtures under high pressure, relevant for process engineering and materials science (Du Plessis et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.761 - 0.765
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
87140172456589926
assay

≥99.0% (GC)

assay

≥99.0% (GC)

assay

≥99%

assay

-

solubility

<0.0001 g/L

solubility

-

solubility

-

solubility

-

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

-

mp

6 °C

mp

5.5 °C (lit.)

mp

5.5 °C (lit.)

mp

38-40 °C (lit.)

form

liquid

form

liquid

form

-

form

solid

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under n-Tetradecane


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

supp_hazards

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters