Skip to Content
Merck

8.14713

Tetraethylene pentamine

(mixture of aliphatic amines) for synthesis

Synonym(s):

Tetraethylene pentamine, 1,4,7,10,13-Pentaazatridecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H23N5
CAS Number:
Molecular Weight:
189.30
UNSPSC Code:
12352116
EC Index Number:
203-986-2
NACRES:
NA.22
MDL number:
eCl@ss:
32150312
Assay:
≥95.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<0.1 hPa ( 20 °C)

Quality Segment

assay

≥95.0% (GC)

form

liquid

autoignition temp.

330 °C

potency

3990 mg/kg LD50, oral (Rat), 660 mg/kg LD50, skin (Rabbit)

expl. lim.

0.1-15 % (v/v)

pH

11.8 (20 °C, 20 g/L in H2O)

bp

330 °C/1013 hPa

mp

-30 °C

transition temp

flash point 193 °C

density

0.998 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCNCCN)CCNCCN

InChI

1S/C8H23N5/c9-1-3-11-5-7-13-8-6-12-4-2-10/h11-13H,1-10H2

InChI key

FAGUFWYHJQFNRV-UHFFFAOYSA-N

Application

  • Organic solar cells enhancement: It is used in developing water/alcohol-soluble hyperbranched perylene diimide electron transport layers, significantly improving the efficiency of organic solar cells (Zhou et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Tetraethylene pentamine


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point (°F)

325.4 °F - closed cup

Flash Point (°C)

163 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library