Skip to Content
Merck
All Photos(1)

Key Documents

8.05905

Sigma-Aldrich

3-Chloro-2-methyl-1-propene

for synthesis

Synonym(s):

3-Chloro-2-methyl-1-propene, ß-Methallyl chloride, 3-Chloroisobutylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2C(CH3)=CH2
CAS Number:
Molecular Weight:
90.55
MDL number:
UNSPSC Code:
12352101
EC Index Number:
209-251-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

140 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

540 °C

potency

>10000 mg/kg LD50, skin (Rat)

expl. lim.

2.2-10.4 % (v/v)

mp

-80 °C

transition temp

flash point -18 °C

solubility

0.5 g/L

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCC(=C)C

InChI

1S/C4H7Cl/c1-4(2)3-5/h1,3H2,2H3

InChI key

OHXAOPZTJOUYKM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.925 - 0.927
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service