Skip to Content

8.04311

4′-Hydroxyacetophenone

for synthesis

Synonym(s):

4′-Hydroxyacetophenone, 4-Acetylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
4-(OH)C6H4COCH3
CAS Number:
Molecular Weight:
136.15
UNSPSC Code:
12352115
EC Index Number:
202-802-8
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

form

powder

mp

105-108 °C

transition temp

flash point 166 °C

storage temp.

2-30°C

SMILES string

Oc1ccc(cc1)C(=O)C

InChI

1S/C8H8O2/c1-6(9)7-2-4-8(10)5-3-7/h2-5,10H,1H3

InChI key

TXFPEBPIARQUIG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 4′-Hydroxyacetophenone


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

330.8 °F

flash_point_c

166 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library