Skip to Content
Merck
All Photos(1)

Key Documents

8.01791

Sigma-Aldrich

Chlorobenzene

for synthesis

Synonym(s):

Chlorobenzene, Phenyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-628-5
NACRES:
NA.22
grade:
for synthesis
bp:
132 °C/1013 hPa
vapor pressure:
12 hPa ( 20 °C)

grade

for synthesis

Quality Level

vapor pressure

12 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

590 °C

potency

2000-4000 mg/kg LD50, oral (Rat)

expl. lim.

1.3-11 % (v/v)

dilution

(for synthesis)

pH

7 (20 °C)

bp

132 °C/1013 hPa

mp

-45 °C

transition temp

flash point 27 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorobenzeneis a colorless and flammable aromatic compound. In various industries,chlorobenzene is primarily used as a solvent, a degreasing agent, and as achemical intermediate.

Application

Chlorobenzene is used as a solvent:
  • In the preparation of polyurethane-isophorone diisocyanate microcapsules by interfacial polymerization of polyurethane prepolymer.
  • To prepare new donor molecule LGC-D073 for organic solar cells which consist of a core (carbazole), a π-linker (DPP), andan end unit rhodanine.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.105 - 1.107
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of polyurethane microcapsules with different polyols component for encapsulation of isophorone diisocyanate healing agent
Kardar P
Progress in Organic Coatings, 89, 271-276 (2015)
Carbazole and rhodanine based donor molecule with improved processability for high performance organic photovoltaics
Nho S, et al.
Dyes and Pigments, 151, 272-278 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service