Skip to Content
Merck

Skip To

8.00899

Dimethyl malonate

for synthesis

Synonym(s):

Dimethyl malonate, Malonic acid dimethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH2(COOCH3)2
CAS Number:
Molecular Weight:
132.11
UNSPSC Code:
12352108
EC Index Number:
203-597-8
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.15 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

440 °C

potency

5331 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-17.4 % (v/v)

bp

181-183 °C/1013 hPa

mp

-61.9 °C

transition temp

flash point 90 °C

density

1.15 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CC(=O)OC

InChI

1S/C5H8O4/c1-8-4(6)3-5(7)9-2/h3H2,1-2H3

InChI key

BEPAFCGSDWSTEL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.151 - 1.155
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.0089869236D97754
description

for synthesis

description

for synthesis

description

analytical standard

description

ReagentPlus®, 99%

assay

≥99.0% (GC)

assay

≥98.0% (GC)

assay

≥97%

assay

99%

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

liquid

form

liquid

form

-

form

liquid

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

density

1.15 g/cm3 at 20 °C

density

1.055 g/cm3 at 20 °C

density

1.007 g/mL at 25 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

bp

181-183 °C/1013 hPa

bp

199 °C/1013 hPa

bp

156 °C/20 mmHg (lit.)

bp

199 °C (lit.)


Still not finding the right product?

Explore all of our products under Dimethyl malonate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters