If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
vapor pressure
0.009 hPa ( 25 °C)
Quality Segment
assay
≥99% (GC)
form
solid
autoignition temp.
424 °C
potency
177 mg/kg LD50, oral (Rat), 1141 mg/kg LD50, skin (Rabbit)
pH
5.0-7.0 (20 °C, 50 g/L in H2O)
bp
125 °C/33.3 hPa
mp
84 °C
transition temp
flash point 138 °C
solubility
2040 g/L
density
1.127 g/cm3 at 25 °C
bulk density
500 kg/m3
SMILES string
NC(=O)C=C
InChI
1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)
InChI key
HRPVXLWXLXDGHG-UHFFFAOYSA-N
Analysis Note
Melting range
- lower value:
≥ 83 °C
- upper value: ≤ 86 °C
Identity (IR): passes test
1 of 1
This Item | |||
|---|---|---|---|
| description for synthesis | description certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland | description suitable for electrophoresis, ≥99% (HPLC), powder | description analytical standard |
| assay ≥99% (GC) | assay - | assay ≥99% (HPLC) | assay ≥99.0% (GC) |
| Quality Level 200 | Quality Level 300 | Quality Level 200 | Quality Level 100 |
| solubility 2040 g/L | solubility - | solubility water: soluble 2.5 g/10 mL, clear, colorless | solubility water: soluble 200 g/L at 20 °C |
| density 1.127 g/cm3 at 25 °C | density - | density - | density - |
| bp 125 °C/33.3 hPa | bp 125 °C/25 mmHg (lit.) | bp 125 °C/25 mmHg (lit.) | bp 125 °C/25 mmHg (lit.) |
| mp 84 °C | mp 82-86 °C (lit.) | mp 82-86 °C (lit.) | mp 82-86 °C (lit.) |
Still not finding the right product?
Explore all of our products under Acrylamide
signalword
Danger
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral
target_organs
Peripheral nervous system
wgk
WGK 3
flash_point_f
280.4 °F - closed cup
flash_point_c
138 °C - closed cup
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
안녕하세요 제품 8.00830Acrylamide가 제품 A9099와 A3553와 비슷한지 궁금합니다. 차이점이 있나요?. 제품 8.00830는 A9099 및 A3553와 유사한 방식으로 사용할 수 있습니까? "폴리아크릴아미드 젤은 아크릴아미드와 비스-아크릴아미드를 사용하여 준비됩니다" 또한 제품 146072 N,N′-Methylenebis(acrylamide) 와 M7279 사이에 차이점이 있습니까? 답장을 보내 주셔서 감사합니다.
1 answer-
These products are very comparable in terms of quality. All offer a minimum purity specification of 99%. They have the same CAS number and molecular weight. The differences are primarily in the testing and demonstrated suitability. Product 8.00830 is simply tested for purity, melting point, and identity by IR. Product A3553 and product A9909 are tested for free carboxylate, purity, heavy metals (including Fe and Mg) solubility, and conductivity. In addition, product A9909 is also tested for RNAse, DNAse, Protease, and Exonuclease. Finally, product A3553 is tested for suitability in electrophoresis.
Please see the link below to compare the features and benefits of each item:
https://www.sigmaaldrich.com/product/compare?term=800830&focus=products&page=1&perpage=30&sort=relevance&type=product&products=800830%3AMM%3Asial%2CA9099%3ASIGMA%3Asial%2CA3553%3ASIGMA%3AsialProduct 146072 and M7279 are similar. Product 146072 is a research grade suitable for gel preparation. Product M7279 was tested for molecular biology. There is more testing done for item M7279. Please view the sample Certificate of Analysis for each item:
https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/146072-BULKMKCT3390.pdf
https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/M7279-250G0000238507.pdfHelpful?
-



