Skip to Content
Merck
All Photos(1)

Key Documents

7990-OP

Millipore

Sodium dodecyl sulfate solution

OmniPur®, 20%

Synonym(s):

Sodium Lauryl Sulfate, SDS, n-dodecyl sulfate sodium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161902
NACRES:
NA.28

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

ambient

General description

Molecular Biology Grade.

A260 (3%, water): ≤0.1 AU
A280 (3%, water): ≤0.1 AU
Assay (C-12 content): ≥99%
Conductivity: 21900-25500µmhos
DNAse: none detected
pH at 25°C: 5.0-8.0
Phosphate (PO4): ≤0.0005%
RNAse: none detected






Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

  • Examination of surface adsorption of sodium chloride and sodium dodecyl sulfate by surface potential measurement at the air/solution interface.: This research investigates the surface adsorption characteristics of SDS and sodium chloride at the air/solution interface using surface potential measurements. The study provides insights into the molecular interactions at the interface, important for understanding surfactant behavior in biochemical applications (Nakahara et al., 2005).

  • Cross-linking studies on the conformation and dimerization of myelin basic protein in solution.: This research utilizes SDS to study the conformation and dimerization of myelin basic protein, providing insights into protein interactions and stability in biochemical solutions (Golds & Braun, 1978).

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service