Skip to Content

79133BC

GDS PickPen® PIPETMAX® Head Stand

BioControl Systems, for use with GDS PickPen® PIPETMAX System

Synonym(s):

GDS PPMX Head Holder, GDS PPMX Head Mount, GDS PPMX Head Rack, GDS PPMX Head Stand, GDS PPMX Head Support

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121521
NACRES:
NC.81
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen® PIPETMAX System

storage temp.

room temp

General description

GDS PickPen® PIPETMAX® (PPMX) Head Stand is an accessory of the GDS PPMX system. The headstand is made of stainless steel for efficient and long-term use. The stand is designed to hold one PPMX pipette or PickPen® head when not used on the instrument.

Application

GDS PickPen® PIPETMAX® Head stand is used with the GDS PickPen® PIPETMAX® system to complete sample preparation for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library