Skip to Content
Merck

78088BC

MVP ICON® pH/Temperature Probe

BioControl, pH Monitoring for HACCP Management

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.79
UNSPSC Code:
41122409
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MVP ICON®

packaging

pkg of 1 ea

manufacturer/tradename

MVP ICON®

application(s)

food and beverages

compatibility

for use with MVP ICON instrument

storage temp.

room temp

General description

The MVP ICON pH / Temperature Probe is a semi-conductor designed for general purpose pH measurement of liquids or semi-solids ranging from 0 – 14 pH. Temperature measurements, ranging from 0 – 80 °C, may be taken
simultaneously.

Application

Food Processing, HACCP Monitoring, Sanitation
The monitoring of pH levels during the process of food production is a vital step in the production of high-quality foods. Proper pH not only affects the aesthetic and quality of the product but maintaining a proper pH level is also critical for food safety. The ability to measure pH and temperature simultaneously allows for increased efficiency in the plant.
pH, Temperature

Biochem/physiol Actions

The probe features a silver / silver chloride / potassium chloride reference system and a thermistor with automatic temperature compensation. Solid-state componentry built into the MVP ICON instrument controls this sensing element.

Features and Benefits

Probe Technology
The MVP ICON pH / Temperature Probe features an Ion Sensitive Field Effect Transistor (ISFET) solid-state pH sensor – the most powerful pH testing technology available.

Increased Efficiency
The probe measures pH and temperature simultaneously for increased efficiency. Advanced algorithms allow for automatic temperature compensation of pH readings across the entire range.

Durable
The rugged, non-glass design allows the probe to be used throughout the entire plant and in the laboratory.

Other Notes

  • Windows 7 or Windows 10
  • Minimum memory requirement 512 MB; recommend at least 1GB or more of RAM
  • At least 16 GB of hard disk space
  • WXGA video resolution of 1280 x 800 or better
  • Mouse or other pointing device
  • One available USB port
  • Adobe Acrobat Reader or similar PDF Viewer
  • Printer (optional)
Clean probe thoroughly with water, a soft bristled brush (included), and a mild detergent solution. Rinse with deionized water. Leave 1 - 2 drops of deionized water in the black protective cap or clear protective tube, then cover probe tip to prevent dehydration of the reference electrode and allow for faster start-up times.
One MVP ICON pH/Temperature Probe

Legal Information

MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library