Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
manufacturer/tradename
Assurance GDS®, BioControl Systems 73063
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
for use with GDS Rotor-Gene
storage temp.
room temp
General description
NOTE: The vortex mixer is 115V and comes with the US plug only. If located in the EU, please additionally order 73060BC for the European power converter and plug.
Application
Other Notes
- Vortex Mixer
- Microtiter plate attachment
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description Particle mixer for use with Assurance® GDS PCR system | description Immunomagnetic separation device for use with Assurance® GDS PCR system | description BioControl Systems, for use with GDS Rotor-Gene® | description BioControl, Molecular based PCR test for the detection of E. coli O157:H7 in food and environmental samples |
| compatibility for use with GDS Rotor-Gene | compatibility for use with GDS Rotor-Gene | compatibility for use with GDS Rotor-Gene® | compatibility BioControl GDS, for use with GDS Rotor-Gene |
| storage temp. room temp | storage temp. room temp | storage temp. 2-8°C | storage temp. 2-8°C |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| manufacturer/tradename Assurance GDS®, BioControl Systems 73063 | manufacturer/tradename Assurance GDS®, BioControl Systems 73091 | manufacturer/tradename BioControl Systems | manufacturer/tradename BioControl Systems 71007-576ATM |
| technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



