Skip to Content
Merck
All Photos(1)

Documents

71493-M

Millipore

HRV 3C Protease

Highly efficient fusion tag removal at low temperature

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

concentration

1.8-3.0 unit/μL

shipped in

dry ice
wet ice

storage temp.

−20°C

General description

Recombinant type 14 3C protease from human rhinovirus (HRV 3C) is a recombinant restriction-grade protease. HRV 3C is a highly purified recombinant 6XHis-fusion protein, that recognizes the same cleavage site as the native enzyme: LeuGluValLeuPheGln?GlyPro. The small, 22-kDa size of the protease, optimal activity at 4°C, high specificity, and His Tag® fusion make HRV 3C protease an ideal choice for rapid removal of fusion tags. The pET expression vectors pET-47b(+) through pET-50b(+) incorporate the HRV 3C protease cleavage site in combinations withHis Tag®, S•Tag, thioredoxin (Trx•Tag), glutathione-S-transferase (GST•Tag), and NusA (Nus•Tag) sequences. The combination of pET-47b(+) to pET-50b(+) vectors for expression, HRV 3C protease for fusion tag cleavage, and Ni-NTA His•Bind metal affinity chromatography for protein purification as well as protease and fusion tag removal allows production of recombinant proteins free of vector-encoded sequences. Other expression vectors, including the pET-52b(+) vector, the InsectDirect pIEx-9 vector, and the multisystem pTriEx-6 vector, all incorporate the HRV 3C protease cleavage site in combination with the Strep•Tag II and His•Tag sequences. Supplied as: 1.8-3.0 U/µl in 50 mM Tris-HCl (pH 8.0), 150 mM NaCl, 1 mM EDTA, 0.5 mM THP, and 50% glycerol.

Application

  • One unit will cleave >95% of 100 µg His•Tag fusion control protein in 50 mM Tris-HCl, 150 mM NaCl, pH 7.5 at 4°C for 16 h.

Components

  • 500 U - HRV 3C Protease
  • 10 µg - HRV 3C Cleavage Control Protein
  • 10 ml - 10X HRV 3C Cleavage Buffer

Warning

Toxicity: Standard Handling

Physical form

In 50 mM Tris-HCl (pH 8.0), 150 mM NaCl, 1 mM EDTA, 0.5 mM THP, and 50% glycerol.

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find highly-specific cleavage enzymes for isolating proteins during fusion protein purification including enterokinase, factor Xa, HRV 3C protease, and thrombin.

Find highly-specific cleavage enzymes for isolating proteins during fusion protein purification including enterokinase, factor Xa, HRV 3C protease, and thrombin.

Find highly-specific cleavage enzymes for isolating proteins during fusion protein purification including enterokinase, factor Xa, HRV 3C protease, and thrombin.

Find highly-specific cleavage enzymes for isolating proteins during fusion protein purification including enterokinase, factor Xa, HRV 3C protease, and thrombin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service