71209
(DE3)pLysS Competent Cell Set 1 - Novagen
Escherichia coli, rod shaped
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106202
NACRES:
NA.81
Recommended Products
Product Name
(DE3)pLysS Competent Cell Set 1 - Novagen,
biological source
Escherichia coli
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
shipped in
dry ice
storage temp.
−70°C
Related Categories
General description
This product is covered under license from Brookhaven National Labs. Commercial entities need to obtain a research use license prior to purchase. Please contact your licensing department to confirm that your company already holds a research use license.
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Components
•0.2 mlBL21(DE3)pLysS Competent Cells
•0.2 mlBLR(DE3)pLysS Competent Cells
•0.2 mlHMS174(DE3)pLysS Competent Cells
•0.2 mlTuner(DE3)pLysS Competent Cells
•10 µlTest Plasmid
•4 × 2 mlSOC Medium
•0.2 mlBLR(DE3)pLysS Competent Cells
•0.2 mlHMS174(DE3)pLysS Competent Cells
•0.2 mlTuner(DE3)pLysS Competent Cells
•10 µlTest Plasmid
•4 × 2 mlSOC Medium
Warning
Toxicity: Multiple Toxicity Values, refer to MSDS (O)
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service