Skip to Content
Merck

Skip To

71012

HT96 BL21(DE3) Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

BL21 strain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106202
NACRES:
NA.81
Biological source:
Escherichia coli
Growth mode:
adherent or suspension
Morphology:
rod shaped
Manufacturer/tradename:
Novagen®
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

HT96 BL21(DE3) Competent Cells - Novagen, The HT96 BL21(DE3) Competent Cells are designed for high-throughput transformation and protein expression applications.

biological source

Escherichia coli

Quality Segment

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2×106 cfu/μg

shipped in

dry ice

storage temp.

2-8°C

General description

Genotype: F-ompT hsdSB(rB- mB-) gal dcm (DE3)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
The HT96 BL21(DE3) Competent Cells are designed for high-throughput transformation and protein expression applications.
The HT96<TMSYMBOL></TMSYMBOL> BL21(DE3) Competent Cells aredesigned for high-throughput transformation.The cells are predispensed in 20-l volumesin a sturdy 96-well polypropylene plate,which is compatible with a variety of thermalcyclers and water baths. Wells are individuallysealed and have raised rims to prevent cross-contamination.Seals can be easily piercedwith standard pipet tips or removed for access.Strips of caps are also provided for reliablesealing during manipulation and storage.Groups of 24 wells can be easily split fromthe plate for processing smaller numbers of samples.




HT96 BL21(DE3) Competent Cells
BL21 is the most widely used host background and has the advantage of being deficient in both lon (1) and ompT proteases. The designation (DE3) indicates that the strain is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter.

Features and Benefits

• High-throughput, 96-well format

Other Notes

1 plate4 plates20 plates

•1 plate4 plates20 platesHT96 BL21(DE3) Competent Cells

•1 × 14 ml4 × 14 ml20 × 14 mlSOC Medium

•1 × 10 µl2 × 10 µl10 × 10 µlTest Plasmid

•1 pkg4 pkg20 pkg8-Cap Strips

•1420Reagent Reservoirs
Phillips, T.A., et al. 1984. J. Bacteriol.159, 283.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the “EMD Product”) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.
Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
694546905369449
description

The HT96 BL21(DE3) Competent Cells are designed for high-throughput transformation and protein expression applications.

description

HMS174 strains provide high transformation efficiencies and the recA mutation in a K-12 background. T7 lysozyme expression suppresses basal T7 expression.

description

BLR is a recA derivative of BL21 that improves plasmid monomer yields and may help stabilize target plasmids containing repetitive sequences.

description

BL21 host strain is the most widely used host background and has the advantage of being deficient in both lon and ompT proteases.

biological source

Escherichia coli

biological source

Escherichia coli

biological source

Escherichia coli

biological source

Escherichia coli

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

growth mode

adherent or suspension

growth mode

adherent or suspension

growth mode

adherent or suspension

growth mode

adherent or suspension

morphology

rod shaped

morphology

rod shaped

morphology

rod shaped

morphology

rod shaped

storage temp.

2-8°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters