Skip to Content
Merck
All Photos(1)

Documents

70664

Millipore

Benzonase® Nuclease

Purity > 99%, Effective viscosity reduction and removal of nucleic acids from protein solutions

Synonym(s):

Endonuclease from Serratia marcescens

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

product name

Benzonase® Nuclease, Purity > 99%, Effective viscosity reduction and removal of nucleic acids from protein solutions

biological source

Serratia marcescens

Quality Level

recombinant

expressed in E. coli

Assay

>99% (SDS-PAGE)

form

buffered aqueous glycerol solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze

concentration

25-29 units/μL

impurities

<0.25 EU/kU Total endotoxin

application(s)

research use

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Benzonase® Nuclease is a genetically engineered endonuclease from Serratia marcescens. It degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) while having no proteolytic activity. It is effective over a wide range of conditions and possesses an exceptionally high specific activity. The enzyme completely digests nucleic acids to 5′-monophosphateterminated oligonucleotides 2 to 5 bases in length (below the hybridization limit), which is ideal for removal of nucleic acids from recombinant proteins, enabling compliance with FDA guidelines for nucleic acid contamination. The ability of Benzonase to rapidly hydrolyze nucleic acids makes the enzyme an excellent choice for viscosity reduction to reduce processing time and increase yields of protein. For example, the enzyme is compatible with BugBuster and PopCulture Protein Extraction Reagents and can therefore be added along with these reagents to eliminate viscosity and remove nucleic acids from E. coli extracts.

The enzyme consists of two subunits of30 kDa each. It is functional between pH 6 and 10 and from 0-42°C and requires1-2 mM Mg2+ for activation. The enzyme is also active in the presence of ionic and non-ionic detergents, reducing agents, PMSF(1 mM), EDTA (1 mM) and urea (relative activity depends on specific conditions).Activity is inhibited by > 150 mM monovalent cations, > 100 mM phosphate, > 100 mMammonium sulfate, or > 100 mM guanidine HCl.

Benzonase Nuclease is available in ultrapure (> 99% by SDS-PAGE) and pure (> 90%) grades at a standard concentration of 25-29 U/µl and at a high concentration (HC) of 250 U/µl. Both preparations are free of detectable protease and have specific activity> 1 × 106 U/mg protein. The > 99% purity grade is tested for endotoxins and contains< 0.25 EU/1000 units. The product is supplied in 50% glycerol. Store at -20&#176;C.



Total endotoxin:< 0.25 EU/1,000 units. Purity: > 99% by SDS-PAGE.
Effective viscosity reduction and removal of nucleic acids from protein solutions

Application

Used for the removal of nucleic acid from protein samples.

Biochem/physiol Actions

Digests native or heat-denatured DNA and RNA.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that causes a ΔA₂₆₀ of 1.0 in 30 minutes, which corresponds to complete digestion of 37 µg DNA.

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erika C Urdaneta et al.
Methods (San Diego, Calif.), 178, 72-82 (2019-10-07)
Post-transcriptional regulation of gene expression in cells is facilitated by formation of RNA-protein complexes (RNPs). While many methods to study eukaryotic (m)RNPs rely on purification of polyadenylated RNA, other important regulatory RNA classes or bacterial mRNA could not be investigated
Timothy N Audam et al.
American journal of physiology. Heart and circulatory physiology, 319(1), H109-H122 (2020-05-23)
Although cell therapy-mediated cardiac repair offers promise for treatment/management of heart failure, lack of fundamental understanding of how cell therapy works limits its translational potential. In particular, whether reparative cells from failing hearts differ from cells derived from nonfailing hearts
Thach H Chu et al.
PLoS pathogens, 16(2), e1008083-e1008083 (2020-02-25)
Antibody functions such as neutralization require recognition of antigen by the Fab region, while effector functions are additionally mediated by interactions of the Fc region with soluble factors and cellular receptors. The efficacy of individual antibodies varies based on Fab
Kee Wui Huang et al.
Frontiers in cellular neuroscience, 14, 65-65 (2020-04-09)
Viral vectors are essential tools for the study of neural circuits, with glycoprotein-deleted rabies viruses being widely used for monosynaptic retrograde tracing to map connectivity between specific cell types in the nervous system. However, the use of rabies virus is
Gil Mandelbaum et al.
Neuron, 102(3), 636-652 (2019-03-25)
The thalamic parafascicular nucleus (PF), an excitatory input to the basal ganglia, is targeted with deep-brain stimulation to alleviate a range of neuropsychiatric symptoms. Furthermore, PF lesions disrupt the execution of correct motor actions in uncertain environments. Nevertheless, the circuitry

Articles

Benzonase®endonuclease efficiently removes nucleic acid contaminants from viral production, crucial for cell and gene therapies and vaccines.

Benzonase®endonuclease efficiently removes nucleic acid contaminants from viral production, crucial for cell and gene therapies and vaccines.

Benzonase®endonuclease efficiently removes nucleic acid contaminants from viral production, crucial for cell and gene therapies and vaccines.

Benzonase®endonuclease efficiently removes nucleic acid contaminants from viral production, crucial for cell and gene therapies and vaccines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service