Skip to Content
Merck

692000

ZM 336372

A potent, cell-permeable, reversible, ATP-competitive and specific inhibitor of the protein kinase c-Raf (IC50 = 70 nM).

Synonym(s):

ZM 336372, N-[5-(3-Dimethylaminobenzamido)-2-methylphenyl]-4-hydroxybenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C23H23N3O3
CAS Number:
Molecular Weight:
389.45
UNSPSC Code:
12352117
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

ZM 336372, A potent, cell-permeable, reversible, ATP-competitive and specific inhibitor of the protein kinase c-Raf (IC50 = 70 nM).

SMILES string

N(C)(C)c1cc(ccc1)C(=O)Nc2cc(c(cc2)C)NC(=O)c3ccc(cc3)O

InChI

1S/C23H23N3O3/c1-15-7-10-18(24-23(29)17-5-4-6-19(13-17)26(2)3)14-21(15)25-22(28)16-8-11-20(27)12-9-16/h4-14,27H,1-3H3,(H,24,29)(H,25,28)

InChI key

PYEFPDQFAZNXLI-UHFFFAOYSA-N

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

Quality Level

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Protein kinase c-Raf
Product competes with ATP.
Reversible: yes
Target IC50: 70 nM against protein kinase c-Raf

Disclaimer

Toxicity: Standard Handling (A)

General description

A potent, cell-permeable, reversible, ATP-competitive and specific inhibitor of the protein kinase c-Raf (IC50 = 70 nM). Inhibits c-Raf with ten-fold increased potency compared to B-Raf, but does not inhibit many other protein kinases (even at 50 µM) with the exception of SAPK2a/p38α (IC50 = 2 µM) and SAPK2b/p38β2 (IC50 = 2 µM). The inhibition of c-Raf by ZM 336372 is competitive with respect to ATP.

Legal Information

Sold under license of PCT Application W098/22,103.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Other Notes

Hall-Jackson, C.A., et al. 1999. Chem. Biol.6, 559.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 3 months at -20°C.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Human Kinome Poster: The InhibitorSelect™ Protein Kinase Inhibitor Libraries provide broad coverage of the human kinome as shown here. The depicted human kinome dendrogram of 518 kinases are classified into five broad groups, 90 families, and 145 subfamilies. Inhibitor coverage was assigned based upon published data related to potency (IC50, EC50, Kd, etc.) for individual kinases harvested from the literature. Colored dots denote which library contains an inhibitor with demonstrated potent activity against the designated kinase and do not necessarily reflect known specificity of the inhibitor. Coverage of lipid and atypical kinases are depicted as a separate dendrogram. As shown, Calbiochem® Protein Kinase Inhibitor Libraries cover all major kinase families including TK, CMGC, CAMK, AGC, CK1, STE, TKL, as well as Lipid or Atypical kinase families.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service