681535
Withaferin A
from Withania somnifera, ≥94% (HPLC), solid, anti-inflammatory lactone, Calbiochem®
Synonym(s):
Withaferin A, Withania somnifera
About This Item
Recommended Products
Product Name
Withaferin A, Withania somnifera, A cell-permeable steroidal lactone that displays antitumor, anti-inflammatory, radiosensitizing, and immunosuppressive properties.
Quality Level
description
RTECS - KE7288500
Assay
≥94% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 10 mg/mL
ethanol: 5 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
O1[C@H](CC(=C(C1=O)CO)C)[C@H]([C@@H]2[C@@]3([C@H]([C@H]4[C@@H]([C@@]5([C@@]6(O[C@@H]6C4)[C@H](C=CC5=O)O)C)CC3)CC2)C)C
InChI
1S/C28H38O6/c1-14-11-21(33-25(32)17(14)13-29)15(2)18-5-6-19-16-12-24-28(34-24)23(31)8-7-22(30)27(28,4)20(16)9-10-26(18,19)3/h7-8,15-16,18-21,23-24,29,31H,5-6,9-13H2,1-4H3/t15-,16-,18+,19-,20-,21+,23-,24+,26+,27-,28+/m0/s1
InChI key
DBRXOUCRJQVYJQ-CKNDUULBSA-N
General description
Biochem/physiol Actions
A steroidal lactone that displays antitumor, anti-inflammatory, radiosensitizing, and immunosuppressive properties.
Packaging
Warning
Other Notes
Jeyaprakasam, B., et al. 2003. Life Sci.74, 125.
Devi, P.U., et al. 1995. Cancer Lett.95, 189.
Kupchan, S.M., et al. 1969. J. Org. Chem.34, 3858.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
438.8 °F
Flash Point(C)
226 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service