Skip to Content
Merck

Skip To

676380

Valproic Acid sodium salt

≥98% (HPLC), solid, HDAC1 inhibitor, Calbiochem®

Synonym(s):

Valproic Acid, Sodium Salt, 2-Propylpentanoic Acid, Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H15NaO2
CAS Number:
Molecular Weight:
166.19
UNSPSC Code:
12352106
NACRES:
NA.77
MDL number:
eCl@ss:
42020819
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Valproic Acid, Sodium Salt, A cell-permeable, short-chained fatty acid that inhibits histone deacetylase (IC50 = 400 µM for HDAC1).

Quality Segment

description

Merck USA index - 14, 9913

assay

≥98% (HPLC)

form

solid

potency

400 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)C(CCC)CCC

InChI

1S/C8H16O2.Na/c1-3-5-7(6-4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1

InChI key

AEQFSUDEHCCHBT-UHFFFAOYSA-M

General description

A cell-permeable, short-chained fatty acid that inhibits histone deacetylase activity (IC50 = 400 µM for HDAC1). Induces differentiation and inhibits proliferation of cell lines derived from human malignant gliomas. At therapeutic levels (350 µM-1.04 mM), causes inositol depletion, inhibits both GSK-3α and -3β, activates the ERK pathway, and produces neurotropic effects. Has been used as an anti-epileptic agent. Also reported to stimulate peroxisome proliferator-activated receptor (PPAR) activity. Displays a potent teratogenic activity in humans and rodent models.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HDAC1
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Other Notes

Gottlicher, M., et al. 2001. EMBO J.20, 6969.
Knupfer, M.M., et al. 2001. Anticancer Res.21, 347.
Phiel, C.J., et al. 2001. J. Biol. Chem.276, 36734.
Vaden, D.L., et al. 2001. J. Biol. Chem.276, 15466.
Yuan, P.X., et al. 2001. J. Biol. Chem.276, 31674.
Chen, G., et al. 1999. J. Neurochem.72, 1327.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P4543524403459620
form

solid

form

powder

form

crystalline solid

form

solid

assay

≥98% (HPLC)

assay

98%

assay

≥98% (HPLC)

assay

≥98% (TLC)

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

−20°C

storage condition

OK to freeze, desiccated (hygroscopic)

storage condition

-

storage condition

OK to freeze, desiccated (hygroscopic)

storage condition

OK to freeze, protect from light


Still not finding the right product?

Explore all of our products under Valproic Acid, Sodium Salt


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters