Skip to Content
Merck
All Photos(2)

Key Documents

616520

Sigma-Aldrich

TRPV4 Antagonist I, RN-1734

The TRPV4 Antagonist I, RN-1734, also referenced under CAS 946387-07-1, controls the biological activity of TRPV4. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

TRPV4 Antagonist I, RN-1734, 2,4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide, Transient Receptor Potential Vanilloid-4 Antagonist I, RN1734, RN1734, 2,4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide, Transient Receptor Potential Vanilloid-4 Antagonist I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22Cl2N2O2S
CAS Number:
Molecular Weight:
353.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N(C(C)C)CCNC(C)C)c1c(cc(cc1)Cl)Cl

InChI

1S/C14H22Cl2N2O2S/c1-10(2)17-7-8-18(11(3)4)21(19,20)14-6-5-12(15)9-13(14)16/h5-6,9-11,17H,7-8H2,1-4H3

InChI key

IHYZMEAZAIFMTN-UHFFFAOYSA-N

General description

A cell-permeable benzenesulfonamide compound that selectively antagonizes TRPV4 agonist 4α-PDD- (Cat. No. 524394) stimulated Ca2+ influx (IC50 = 2.3, 3.2 and 5.9 µM, respectively, in HEK293 expressing human, rat, or mouse TRPV4) without affecting the activity of three other human TRP (transient receptor potential) channels, including capsaicin- (Cat. No. 211275) activated TRPV1, camphor-activated TRPV3, and methanol-activated TRPM8 (IC50 >30 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Vincent, F., et al. 2009. Biochem. Biophys. Res. Commun.389, 490.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service