Skip to Content
Merck
All Photos(1)

Key Documents

598101

Sigma-Aldrich

Thioredoxin, Human, Recombinant, E. coli

Synonym(s):

Thioredoxin, Human, Recombinant, E. coli, rhTRX

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

description

Merck USA index - 14, 9358

Assay

≥90% (SDS-PAGE)

form

lyophilized solid

specific activity

≥1.7 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human thioredoxin expressed in E. coli. A ubiquitous protein with two redox-active half-cysteine residues that function as a general disulfide reductant. Works efficiently on model compounds (e.g. Ellman′s reagent, Cat. No. 322123) and protein disulfides. Plays an essential role in cellular protection against oxidative stress and cell death. Inhibits apoptosis in Jurkat T-cells and human PBL blasts. Occurs as a single polypeptide chain.
Recombinant, human thioredoxin expressed in E. coli. A ubiquitous protein with two redox-active half-cysteine residues. Functions as a general disulfide reductant. Works efficiently on model compounds (e.g. Ellman’s reagent, Cat. No. 322123) and protein disulfides. Plays an essential role in cellular protection against oxidative stress and cell death. Inhibits apoptosis in Jurkat T cells and human PBL.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit (insulin reduction unit) is calculated by measuring the change in absorbance at 650 nm with 330 µM DTT as the reductant at 25°C, pH 6.5.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Reconstitute in physiological buffer containing 3 mM EDTA and 1-5 mM DTT.

Other Notes

Noda, N., et al. 1997. Nippon Rinsho 55, 461.
Wollman, E.E., et al. 1997. Biochem. Biophys. Res. Commun. 230, 602.
Masutani, H., et al. 1996. Immunol. Lett. 54, 67.
Saito, I., et al. 1996. Arthritis Rheumatism, 39, 773.
Saito, N., et al. 1995. J. Immunol. 154, 3194.
Buchanon, B.B., et al. 1994. Arch. Biochem. Biophys. 314, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service