Skip to Content
Merck
All Photos(1)

Key Documents

557360

Sigma-Aldrich

Roscovitine

≥95% (HPLC), solid, Cdk inhibitor, Calbiochem®

Synonym(s):

Roscovitine, 2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-benzylamino-9-isopropylpurine, Cdk7 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26N6O
CAS Number:
Molecular Weight:
354.45
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Roscovitine, A potent, reversible, and selective inhibitor of Cdks that exhibits about 10-fold greater efficacy towards p34-cdk1 and p33-cdk2 and 20-fold greater efficacy towards p33-cdk5 relative to Olomoucine.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]1(c2nc(nc(c2nc1)NCc3ccccc3)N[C@@H](CO)CC)C(C)C

InChI

1S/C19H26N6O/c1-4-15(11-26)22-19-23-17(20-10-14-8-6-5-7-9-14)16-18(24-19)25(12-21-16)13(2)3/h5-9,12-13,15,26H,4,10-11H2,1-3H3,(H2,20,22,23,24)/t15-/m1/s1

InChI key

BTIHMVBBUGXLCJ-OAHLLOKOSA-N

General description

A potent, reversible, and selective inhibitor of cyclin-dependent kinases (Cdks) that exhibits about ten-fold greater efficacy towards p34cdk1 and p33cdk2 and twenty-fold greater efficacy towards p33cdk5 relative to Olomoucine (Cat. No. 495620). Inhibits p34cdk1/cyclin B (IC50 = 650 nM), p33cdk2/cyclin A (IC50 = 700 nM), p33cdk2/cyclin E (IC50 = 700 nM), and p33cdk5/p35 (IC50 = 200 nM) by competing for the ATP binding domain of the kinases. Exhibits reduced sensitivity towards related kinases including ERK1 and ERK2 (IC50 = 34 µM and 14 µM, respectively). Roscovitine does not significantly affect the activity of other protein kinases even at 100 µM, including p34cdk4/cyclin D1 and p40cdk6/cyclin D2. Compared to olomoucine, roscovitine displays increased anti-mitotic activity at the G1/S and G2/M phases of the cell cycle. A 50 mM (5 mg/282 µl) solution of Roscovitine (Cat. No. 557364) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: no
Primary Target
p34cdk1/cyclin B
Product does not compete with ATP.
Reversible: yes
Target IC50: 650 nM, 700 nM, 700 nM, 200 nM, against p34cdk1/cyclin B, p33cdk2/cyclin A, p33cdk2/cyclin E, p33cdk5/p35, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for at least 6 months at -20°C.

Other Notes

De Azevedo, W.F., et al. 1997. Eur. J. Biochem.243, 518.
Meijer, L., et al. 1997. Eur. J. Biochem.243, 527.
Meijer, L., et al. 1996. Trends Cell Biol. 6, 393.
Rudolph, B., et al. 1996. EMBO J. 15, 3053.

Legal Information

Sold under license of U.S. Patent 6,316,456
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arzu Ulu et al.
Cellular signalling, 80, 109926-109926 (2021-01-20)
The Neuroepithelial transforming gene 1 (Net1) is a RhoA subfamily guanine nucleotide exchange factor that is overexpressed in a number of cancers and contributes to cancer cell motility and proliferation. Net1 also plays a Rho GTPase independent role in mitotic
Taichi Igarashi et al.
Nature communications, 14(1), 4991-4991 (2023-08-18)
Activation of the KRAS oncogene is a source of replication stress, but how this stress is generated and how it is tolerated by cancer cells remain poorly understood. Here we show that induction of KRASG12V expression in untransformed cells triggers
A-Ching Chao et al.
Biochimica et biophysica acta. Molecular cell research, 1867(4), 118628-118628 (2019-12-31)
One neurotoxic mechanism of amyloid-beta peptide (Aβ), the major component of senile plaques in the brains of Alzheimer's disease (AD) patients, is to trigger cell cycle reentry in fully differentiated neurons. However, the detailed underlying mechanisms remain unclear. Using Aβ25-35-treated
Sandipan Ray et al.
Life science alliance, 2(6) (2019-12-04)
Determining the exact targets and mechanisms of action of drug molecules that modulate circadian rhythms is critical to develop novel compounds to treat clock-related disorders. Here, we have used phenotypic proteomic profiling (PPP) to systematically determine molecular targets of four
Christiana Kontaxi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 43(11), 2002-2020 (2023-02-10)
Cyclin-dependent kinase-like 5 (CDKL5) deficiency disorder (CDD) is a severe early-onset epileptic encephalopathy resulting mainly from de novo mutations in the X-linked CDKL5 gene. To determine whether loss of presynaptic CDKL5 function contributes to CDD, we examined synaptic vesicle (SV)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service