524739
Phosphodiesterase 10A2, His•Tag®, Human, Recombinant, S. frugiperda
Synonym(s):
PDE10A2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
recombinant
expressed in baculovirus infected Sf21 cells
Quality Level
form
liquid
specific activity
≥0.25 U/μL
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Full-length, recombinant, human, phosphodiesterdase 10A2 (accession no. 10A2) fused at the N-terminus to a His•Tag7reg; sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. Supplied as a partially purified preparation of catalytically active enzyme.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme required to convert 1 µmol cAMP to AMP per min at 30°C, pH 7.5.
Physical form
In RIPA buffer (150 mM NaCl, 10 mM Tris-HCl, 1% NP-40), pH 8.3.
Other Notes
Kotera, J., et. al. 2004. J. Biol. Chem.279, 4366.
Fujishige K., et. al. 1999. Eur. J. Biochem.266, 1118.
Kotera, J., et. al. 1999. Biochem. Biophys. Res. Commun.261, 551.
Fujishige K., et. al. 1999. Eur. J. Biochem.266, 1118.
Kotera, J., et. al. 1999. Biochem. Biophys. Res. Commun.261, 551.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service