Skip to Content
Merck

5.08160

DPDPE

Synonym(s):

DPDPE, δ Opioid Receptor Agonist, DPDPE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H39N5O7S2
CAS Number:
Molecular Weight:
645.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Form:
solid
Assay:
≥99% (HPLC)
Solubility:
water: 1 mg/mL
Color:
off-white
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (HPLC)

Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

water: 1 mg/mL

storage temp.

−20°C

SMILES string

S1SC([C@H](C(=O)NCC(=O)N[C@H](C(=O)N[C@H](C1(C)C)C(=O)O)Cc3ccccc3)NC(=O)[C@@H](N)Cc2ccc(cc2)O)(C)C

InChI

1S/C30H39N5O7S2/c1-29(2)23(34-25(38)20(31)14-18-10-12-19(36)13-11-18)27(40)32-16-22(37)33-21(15-17-8-6-5-7-9-17)26(39)35-24(28(41)42)30(3,4)44-43-29/h5-13,20-21,23-24,36H,14-16,31H2,1-4H3,(H,32,40)(H,33,37)(H,34,38)(H,35,39)(H,41,42)/t20-,21-,23-,24-/m0/s1

InChI key

MCMMCRYPQBNCPH-WMIMKTLMSA-N

General description

A potent agonist selective for δ opioid receptor (EC50 = 5.2 nM). Exhibits anti-convulsant effects in vivo in rats, also used in nociceptive studies.

Biochem/physiol Actions

Primary Target
δ opioid receptor

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, C., 1997. J. Pharmacol. Exp. Ther.283, 1151.
Chandrakumar, S., et al., 1992. J. Med. Chem.35, 2928.
Tortella, F. 1988. Peptides.9, 1177.
Clark, A., et al., 1986. Eur. J. Pharmacol.128, 303.
Tyr-D-Pen-Gly-Phe-D-Pen (2-5 disulfide bond)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library