Skip to Content
Merck

487500

p-Nitrophenyl-β-D-glucuronide

≥98% (HPLC), Chromogenic β-glucuronidase substrate, solid

Synonym(s):

p-Nitrophenyl-β-D-glucuronide, p-Nitrophenyl-β-D-glucopyranosiduronic Acid

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C12H13NO9
CAS Number:
Molecular Weight:
315.23
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

p-Nitrophenyl-β-D-glucuronide, Chromogenic substrate for β-glucuronidase (Km = ~0.22 mM).

SMILES string

[N+](=O)([O-])c1ccc(cc1)O[C@@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)O)O)C(=O)[O-]

InChI

1S/C12H13NO9/c14-7-8(15)10(11(17)18)22-12(9(7)16)21-6-3-1-5(2-4-6)13(19)20/h1-4,7-10,12,14-16H,(H,17,18)/p-1/t7-,8-,9+,10-,12+/m0/s1

InChI key

QSUILVWOWLUOEU-GOVZDWNOSA-M

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

impurities

≤0.1% Free p-Nitrophenol

color

off-white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

Quality Level

Related Categories

Disclaimer

Toxicity: Standard Handling (A)

General description

Chromogenic substrate for β-glucuronidase (Km = ~0.22 mM).

Other Notes

Mariscal, A., et al. 1998. Arch. Environ. Contam. Toxicol. 35, 588.
Kim, D. H., et al. 1995. Biol. Pharm. Bull.18, 1184.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service