Skip to Content
Merck
All Photos(1)

Key Documents

48-680MAG

Millipore

MILLIPLEX MAP Multi-Pathway Magnetic Bead 9-Plex - Cell Signaling Multiplex Assay

Synonym(s):

ERK Cell Signaling Assay, Luminex® Cell Signaling Assay, Millipore Cell Signaling Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

storage temp.

2-8°C

General description

The Milliplex® MAP 9-plex Multi-Pathway Signaling Magnetic Bead kit, phosphoprotein, is used to detect changes in phosphorylated ERK/MAP kinase 1/2 (Thr185/Tyr187), Akt (Ser473), STAT3 (Ser727), JNK (Thr183/Tyr185), p70S6K (Thr412), NFkB (Ser536), STAT5A/B (Tyr694/699), CREB (Ser133), and p38 (Thr180/Tyr182) in cell lysates using the Luminex® xMAP® technology. The detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures. Each kit has sufficient reagents for one 96-well plate assay.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Analyze a cell signaling panel using this kit. The analytes available for the cell signaling multiplex assay kit are: CREB (pS133), ERK (pT185/pY187), NFκB (pS536), JNK (pT183/pY185), p38 (Thr180/Tyr182), p70S6K (Thr412), STAT3 (pS727), STAT5A/B (pY694/699), Akt (pS473).
Research Category
Cell Signaling
Research Sub Category
Oncology
Used to detect/quantify:CREB (pS133)|ERK (pT185/pY187)|NFκB (pS536)|JNK (pT183/pY185)|p38 (Thr180/Tyr182)|p70S6K (Thr412)|STAT3 (pS727)|STAT5A/B (pY694/699)|Akt (pS473)

Storage and Stability

Recommended storage for kit components is 2 - 8°C

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher P Stanley et al.
Pharmacological research, 113(Pt A), 356-363 (2016-10-21)
The endocannabinoid anandamide (AEA) causes vasorelaxation in animal studies. Although circulating AEA levels are increased in many pathologies, little is known about its vascular effects in humans. The aim of this work was to characterise the effects of AEA in
Lee A Denson et al.
Inflammatory bowel diseases, 25(3), 547-560 (2018-08-21)
Granulocyte-macrophage colony-stimulating factor auto-antibodies (GMAbs) suppress neutrophil-extrinsic GM-CSF signaling and increase risk for stricturing behavior in Crohn's disease (CD). We aimed to define clinical, genomic, and functional associations with neutrophil-intrinsic GM-CSF signaling. Missense mutations in CSF2RA, CSF2RB, JAK2, STAT5A, and
Fernanda Ursoli Ferreira et al.
International journal of molecular sciences, 20(3) (2019-01-27)
The endothelial-to-mesenchymal transition (EndMT) is a biological process where endothelial cells (ECs) acquire a fibroblastic phenotype after concomitant loss of the apical-basal polarity and intercellular junction proteins. This process is critical to embryonic development and is involved in diseases such
Anna Coulibaly et al.
Scientific reports, 11(1), 7023-7023 (2021-03-31)
Natural killer (NK) cells mediate innate host defense against microbial infection and cancer. Hypoxia and low glucose are characteristic for these tissue lesions but do not affect early interferon (IFN) γ and CC chemokine release by interleukin 15 (IL-15) primed
Sophie A Millar et al.
Journal of cellular physiology, 235(4), 3414-3424 (2019-09-25)
Some human observational studies have suggested an anti-inflammatory role of osteocalcin (OCN). An inflammatory protocol using interferon-γ and tumor necrosis factor-α (10 ng/ml) was employed to examine the acute (24 hr) and chronic (144 hr) effects of uncarboxylated OCN (ucOCN) in commercial, primary

Related Content

Learn how multiplexing with MILLIPLEX® multiplex assays can help simplify cancer research by measuring multiple circulating biomarkers at once. Find multiplex panels to analyze JAK-STAT, mTOR, MAPK, SAPK, TGFβ signaling pathways, and more.

Learn how multiplexing with MILLIPLEX® multiplex assays can help simplify cancer research by measuring multiple circulating biomarkers at once. Find multiplex panels to analyze JAK-STAT, mTOR, MAPK, SAPK, TGFβ signaling pathways, and more.

Learn how multiplexing with MILLIPLEX® multiplex assays can help simplify cancer research by measuring multiple circulating biomarkers at once. Find multiplex panels to analyze JAK-STAT, mTOR, MAPK, SAPK, TGFβ signaling pathways, and more.

Learn how multiplexing with MILLIPLEX® multiplex assays can help simplify cancer research by measuring multiple circulating biomarkers at once. Find multiplex panels to analyze JAK-STAT, mTOR, MAPK, SAPK, TGFβ signaling pathways, and more.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service