474788
MG-132
≥95% (HPLC), liquid, proteasome inhibitor, Calbiochem®
Synonym(s):
InSolution MG-132 in EtOH, ≥95% by HPLC, Proteasome Inhibitor XI
About This Item
product name
MG-132 in EtOH, ≥95% by HPLC, InSolution, ≥95%, reversible proteasome inhibitor
Quality Level
Assay
≥95% (HPLC)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
protect from light
shipped in
ambient
storage temp.
−70°C
General description
Biochem/physiol Actions
IKK
Packaging
Warning
Physical form
Reconstitution
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
53.6 °F - closed cup - Information refers to the main ingredient.
Flash Point(C)
12 °C - closed cup - Information refers to the main ingredient.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service