Skip to Content
Merck

4512

CellPrime r Insulin recombinant human insulin, 1 g

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32160801
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

white

Quality Segment

type

for cell culture

form

powder or crystals

shelf life

48 mo. (at -20 °C.)

loss

<10% loss on drying (w/w)

General description

CellPrime® r Insulin is a non-animal origin, recombinant supplement that provides excellent quality, performance and lot-to-lot consistency. Used in the production of mAbs, viral vaccines and a wide spectrum of other recombinant biomolecules, CellPrime® r Insulin is produced by microbial expression as a recombinant human insulin precursor in E.coli (K12). It is followed by enzymatic cleavage and subsequent purification steps, yielding a crystalline powder product.

Preparation Note

Shipping and storage at -20°C

Legal Information

CellPrime is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For cell culture supplement use only. For further manufacturing use only. Not for human or therapeutic use.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library