Skip to Content
Merck

444151

Nutlin-3

InSolution, ≥98%, Racemic, potent and selective MDM2 antagonist

Synonym(s):

InSolution Nutlin-3, Racemic, MDM2 Inhibitor IV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H30Cl2N4O4
Molecular Weight:
581.49
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
liquid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, potent, and selective MDM2 antagonist (IC50 = 90 nM for Nutlin-3a and 13.6 µM for Nutlin-3b) that displays antitumor properties. Activates the p53 pathway by binding MDM2 in the p53-binding pocket and inhibiting the MDM2-p53 interaction. Shown to induce apoptosis in several cancer cell lines with wild-type p53 and in xenografted tumor mice.

Packaging

Packaged under inert gas

Physical form

A 10 mM (1 mg/172 µl) solution of MDM2 Antagonist, Nutlin-3, Racemic (Cat. No. 444143) in DMSO

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Thompson, T., et al. 2004. J. Biol. Chem.279, 53015.
Vassilev, L.T., et al. 2004. Science303, 844.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Christophe Le Clorennec et al.
International journal of molecular sciences, 24(18) (2023-09-28)
Neuroblastoma (NB) is a pediatric malignancy originating from neural crest cells of the sympathetic nervous system that accounts for 15% of all pediatric cancer deaths. Despite advances in treatment, high-risk NB remains difficult to cure, highlighting the need for novel
B Lamprecht et al.
The British journal of dermatology, 167(2), 240-246 (2012-03-06)
Primary cutaneous T-cell lymphomas (CTCLs) are a heterogeneous group with Sézary syndrome (SS) as one of the most aggressive variants. Recently, we identified a loss of E2A as a recurrent event in SS, which enhanced proliferation via upregulation of the
Anthony M Pettinato et al.
Cell reports, 35(5), 109088-109088 (2021-05-06)
Human cardiac regeneration is limited by low cardiomyocyte replicative rates and progressive polyploidization by unclear mechanisms. To study this process, we engineer a human cardiomyocyte model to track replication and polyploidization using fluorescently tagged cyclin B1 and cardiac troponin T.



Global Trade Item Number

SKUGTIN
444151-1MG04055977205466