Skip to Content
Merck
All Photos(1)

Key Documents

411550

Sigma-Aldrich

Mouse Anti-Human IgG, Fc Fragment Specific (HP6043) Peroxidase Conjugate

liquid, clone HP6043, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

HP6043, monoclonal

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

General description

Purified mouse monoclonal antibody conjugated to horseradish peroxidase. Recognizes human IgG Fc fragment.
This Mouse Anti-Human IgG, Fc Fragment Specific (HP6043) Peroxidase Conjugate is validated for use in Enzyme Immunoassay for the detection of Human IgG, Fc Fragment Specific.

Application

Enzyme Immunoassay (1:1000, see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 10 mg/ml BSA.

Other Notes

Antibody should be titrated for optimal results in individual systems.



Cross-reactivity by ELISA against human myeloma proteins:

Human IgG1: 100%

Human IgG2: 100%

Human IgG3: 100%

Human IgG4: 100%

Human IgM: <0.2%

Human IgA: <0.1%

Human IgE: <0.1%



Cross-reactivity by ELISA against chimeric antibodies:

Human IgG1: 100%

Human IgG2: 100%

Human IgG3: 100%

Human IgG4: 100%

Human IgM: <0.01%

Human IgA: <0.01%

Human IgE: <0.01%



There is no detectable binding to bovine, goat, horse sheep IgG, so these species sera can be used for assay blocking buffers. Modification of Nakane and Kawaoi method used for enzyme coupling.
Hamilton, R.G. 1993. Clin. Chem.39, 1988.
Hamilton, R.G. and Morrison, S.M. 1993. J. Immunolo. Meth.158, 107.
Hamilton, R.G. 1990. Ann. Biol. Clin.48, 473.
Hamilton, R.G., et al. 1988. J. Immunoassay.9, 275.
Hamilton, R.G., et al. 1987. Hybridoma6, 204.
Nakane, P.K. and Kawaoi A., 1974. J. Histochem Cytochem.22, 1084.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farhad Khosravi et al.
Nanotechnology, 27(13), 13LT02-13LT02 (2016-02-24)
We demonstrate the rapid and label-free capture of breast cancer cells spiked in buffy coats using nanotube-antibody micro-arrays. Single wall carbon nanotube arrays were manufactured using photo-lithography, metal deposition, and etching techniques. Anti-epithelial cell adhesion molecule (EpCAM) antibodies were functionalized
Farhad Khosravi et al.
Biosensors, 7(2) (2017-04-20)
This study demonstrates the rapid and label-free detection of Interleukin-6 (IL-6) using carbon nanotube micro-arrays with aptamer as the molecular recognition element. Single wall carbon nanotubes micro-arrays biosensors were manufactured using photo-lithography, metal deposition, and etching techniques. Nanotube biosensors were
Tiina Kelkka et al.
Leukemia, 36(9), 2317-2327 (2022-08-05)
In immune aplastic anemia (IAA), severe pancytopenia results from the immune-mediated destruction of hematopoietic stem cells. Several autoantibodies have been reported, but no clinically applicable autoantibody tests are available for IAA. We screened autoantibodies using a microarray containing >9000 proteins
Farhad Khosravi et al.
Nanotechnology, 27(44), 44LT03-44LT03 (2016-09-30)
We demonstrate the rapid and label-free capture of breast cancer cells spiked in blood using nanotube-antibody micro-arrays. 76-element single wall carbon nanotube arrays were manufactured using photo-lithography, metal deposition, and etching techniques. Anti-epithelial cell adhesion molecule (anti-EpCAM), Anti-human epithelial growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service