Skip to Content

4015

Imidazole

ULTROL® Grade, ≥99% (1H-NMR), useful in pH range 6.2-7.8

Synonym(s):

Imidazole, ULTROL® Grade, 1,3-Diaza-2,4-cyclopentadiene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
UNSPSC Code:
12352005
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - NI3325000
Merck USA index - 14, 4912

Quality Segment

assay

≥99% (1H-NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

7.0

solubility

water: 100 mg/mL

cation traces

heavy metals: <1 ppm

application(s)

advanced drug delivery

shipped in

ambient

storage temp.

15-25°C

SMILES string

[nH]1cncc1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

General description

Superior buffer in the pH range of 6.2-7.8. Has a pKa of 7.0 at 20°C.
Useful in the pH range of 6.2-7.8.

Application


  • Prediction of adsorption performance of ZIF-67 for malachite green based on artificial neural network using L-BFGS algorithm.: Utilizing imidazole-based frameworks, specifically ZIF-67, this study showcases the effectiveness of imidazole structures in adsorbing and removing industrial dyes from water, highlighting environmental applications of imidazole in water treatment processes (Xiong C et al., 2024).

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Corrosive (L)


Still not finding the right product?

Explore all of our products under Imidazole


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

293.0 °F - closed cup

flash_point_c

145 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library