Skip to Content
Merck
All Photos(1)

Key Documents

401481

Sigma-Aldrich

IKK-2 Inhibitor IV

The IKK-2 Inhibitor IV, also referenced under CAS 507475-17-4, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

IKK-2 Inhibitor IV, [5-( p-Fluorophenyl)-2-ureido]thiophene-3-carboxamide, TPCA-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10FN3O2S
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to pale yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)c2[s]c(c(c2)C(=O)N)NC(=O)N

InChI

1S/C12H10FN3O2S/c13-7-3-1-6(2-4-7)9-5-8(10(14)17)11(19-9)16-12(15)18/h1-5H,(H2,14,17)(H3,15,16,18)

InChI key

SAYGKHKXGCPTLX-UHFFFAOYSA-N

General description

A cell-permeable ureidocarboxamido thiophene compound that acts as a potent, reversible, and ATP-competitive inhibitor of IKK-2 (IC50 = 18 nM) with selectivity over IKK-1, JNK and p38 MAPK. Inhibits TNF-α production in human monocytes (IC50 in the range of 0.15-2.5 µM) and blocks IL-8 and IL-6 production by synovial fibroblasts (IC50 = 100 nM). Further reduces paw oedema in rat arthritis model (~100% inhibition at a dose of 30 mg per kg). A 10 mM (500 µg/179 µl) solution of IKK-2 Inhibitor IV (Cat. No. 401484) in DMSO is available.
A cell-permeable, reversible, ATP-competitive, and potent inhibitor of IKK-2 (IC50 = 18 nM) that exhibits selectivity over IKK-1, JNK, and p38 MAPK. Inhibits TNF-α production in human monocytes (IC50 in the range of 0.15-2.5 µM) and blocks IL-8 and IL-6 production by synovial fibroblasts (IC50 = 100 nM). Shown to reduce paw oedema in rat arthritis model (~100% inhibition at a dose of 30 mg per kg).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IKK-2
Product competes with ATP.
Reversible: yes
Target IC50: 18 nM against IKK-2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Podolin, P.L., et al. 2005. J. Pharmacol. Exp. Ther.312, 373.
Karin, M., et al. 2004. Nat. Rev. Drug Discov.3, 17.
Roshak, A.K., et al. 2002. Inflamm. Res.51, S4.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lídia Hernandez et al.
Cancer research, 70(10), 4005-4014 (2010-04-29)
The NF-kappaB family of transcription factors has been implicated in the propagation of ovarian cancer, but the significance of constitutive NF-kappaB signaling in ovarian cancer is unknown. We hypothesized that constitutive NF-kappaB signaling defines a subset of ovarian cancer susceptible
Sang Min Jeon et al.
Virulence, 13(1), 1966-1984 (2022-10-23)
Ohmyungsamycin A (OMS) is a newly identified cyclic peptide that exerts antimicrobial effects against Mycobacterium tuberculosis. However, its role in nontuberculous mycobacteria (NTMs) infections has not been clarified. Mycobacteroides abscessus (Mabc) is a rapidly growing NTM that has emerged as
Haibo Zhang et al.
Science signaling, 17(836), eadd5073-eadd5073 (2024-05-15)
The Ras-mitogen-activated protein kinase (MAPK) pathway is a major target for cancer treatment. To better understand the genetic pathways that modulate cancer cell sensitivity to MAPK pathway inhibitors, we performed a CRISPR knockout screen with MAPK pathway inhibitors on a
Minho Won et al.
Cell death and differentiation, 29(6), 1152-1163 (2021-11-29)
In TNF signaling, ubiquitination of RIP1 functions as an early cell-death checkpoint, which prevents the spatial transition of the signaling complex from complex-I to death-inducing complex-II. Here, we report that ankyrin repeat domain 13a (ANKRD13a) acts as a novel component
Kidong Kang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(3), 4369-4383 (2020-02-07)
In tumor necrosis factor (TNF) signaling, phosphorylation and activation of receptor interacting protein kinase 1 (RIPK1) by upstream kinases is an essential checkpoint in the suppression of TNF-induced cell death. Thus, discovery of pharmacological agents targeting RIPK1 may provide new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service