Skip to Content
Merck

401152

Human IgE, Myeloma

This Human IgE, Myeloma is validated for use as an Immunoassay standard.

Synonym(s):

Human IgE antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
UNSPSC Code:
12352202
Clone:
-
Species reactivity:
-
Application:
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Human myeloma)

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoassay standard)

isotype

IgE

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

General description

Native IgE from human myeloma plasma.
Native IgE from human myeloma plasma. Human immunoglobulin E (IgE) purified from the plasma of a myeloma donor. The isotype of the light chain (kappa or lambda) will vary from batch to batch depending on the starting material.

Application

Immunoassay standard

Physical form

In 150 mM NaCl, 15 mM potassium phosphate buffer, pH 7.4.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).
Prepared from plasma that was shown by certified tests to be nonreactive for HBsAg and for antibodies to HCV, HBc, HIV-1 and HIV-2.

Analysis Note

Single arc by immunoelectrophoresis against antiserum to whole human serum and human IgE

Other Notes

Note: the heavy chain of myeloma IgE may appear as either a single or double band on gel electrophoresis. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Human IgE, Myeloma


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
401152-100UG04055977188899