Skip to Content
Merck

Skip To

346299

D(+)Glucosamine, Hydrochloride

Synonym(s):

D(+)Glucosamine, Hydrochloride, 2-Amino-2-deoxy-D-glucose, Chitosamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · xHCl
CAS Number:
Molecular Weight:
179.17 (free base basis)
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (by Assay)

Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Cl-].NC(C(O)C(O)C(O)CO)C=O.[H+]

InChI

1S/C6H13NO5.ClH/c7-3(1-8)5(11)6(12)4(10)2-9;/h1,3-6,9-12H,2,7H2;1H

InChI key

CBOJBBMQJBVCMW-UHFFFAOYSA-N

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G1514G4875PHR1199
assay

≥99% (by Assay)

assay

≥99%

assay

≥99% (HPLC)

assay

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

300

solubility

water: 5 mg/mL

solubility

water: 100 mg/mL, clear, colorless

solubility

water: 100 mg/mL, clear, colorless

solubility

-

form

solid

form

powder

form

powder

form

-

storage temp.

15-25°C

storage temp.

-

storage temp.

room temp

storage temp.

2-30°C

color

white

color

-

color

white

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the origin (source) for this glucosamine? Is it xeno-free?

    1 answer
    1. While the source of this material may change in the future, currently, this product is derived from crab and/or shrimp shells.

      Helpful?

Reviews

No rating value

Active Filters