Skip to Content
Merck
All Photos(1)

Documents

344930

Sigma-Aldrich

Furin Inhibitor I

≥90% (HPLC), solid, furin inhibitor, Calbiochem®

Synonym(s):

Furin Inhibitor I, Decanoyl-RVKR-CMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H66ClN11O5
Molecular Weight:
744.41
UNSPSC Code:
12352200
NACRES:
NA.28

product name

Furin Inhibitor I, Furin Inhibitor I, is a peptidyl chloromethylketone that binds to the catalytic site of furin and blocks its activity.

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

methanol: 1 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

General description

A peptidyl chloromethylketone that binds irreversibly to the catalytic site of furin and blocks its activity. Hence, it can be used as a high specificity cleavage inhibitor of viral glycoproteins and blocker viral replication. Reported to block the shedding of MT5-MMP by furin and prevent the activation of MT5-MMP. Completely inhibits the cleavage of Boc-RVRR-AMC (50 µM) by purified furin or PACE4. Also inhibits the activity of endothelin converting enzyme and reduces the pro-peptide cleavage of BACE (β-site APP-cleaving enzyme).
A peptidyl chloromethylketone that binds to the catalytic site of furin and blocks its activity. Hence, it can be used as a high specificity cleavage inhibitor of viral glycoproteins and blocker of viral replication. Reported to block the shedding of MT5-MMP by furin and prevent the activation of MT5-MMP. Also shown to reduce the pro-peptide cleavage of BACE (β-site APP-cleaving enzyme).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
viral glycoproteins
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Decanoyl-Arg-Val-Lys-Arg-CMK

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Sugrue, R.J., et al. 2001. J. Gen. Virol.82, 1375.
Wang, X., and Pei, D. 2001. J. Biol. Chem.276, 35953.
Capell, A., et al. 2000. J. Biol. Chem. 275, 30849.
Denault, J.B., et al. 1995. FEBS Lett. 362, 276.
Denault, J.B., et al. 1995. J. Cardiovasc. Pharmacol.26, S47.
Garten, W., et al. 1994. Biochimie 76, 217.
Hallenberger, S., et al. 1992. Nature 360, 358.
Stieneke-Gröber, A., et al. 1992. EMBO J. 11, 2407.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Montalbano Mauro et al.
International journal of molecular sciences, 24(13) (2023-07-14)
Glypican-3 (GPC-3) is a heparin sulfate proteoglycan located extracellularly and anchored to the cell membrane of transformed hepatocytes. GPC-3 is not expressed in normal or cirrhotic liver tissue but is overexpressed in hepatocellular carcinoma (HCC). Because of this, GPC-3 is
Zhujun Ao et al.
iScience, 25(8), 104759-104759 (2022-07-21)
The Delta variant had spread globally in 2021 and caused more serious disease than the original virus and Omicron variant. In this study, we investigated several virological features of Delta spike protein (SPDelta), including protein maturation, its impact on viral
Carlota Oleaga et al.
Journal of lipid research, 62, 100003-100003 (2021-01-12)
Proprotein convertase subtilisin/kexin type 9 (PCSK9) regulates cholesterol metabolism by inducing the degradation of hepatic low density lipoprotein receptors (LDLRs). Plasma PCSK9 has 2 main molecular forms: a 62 kDa mature form (PCSK9_62) and a 55 kDa, furin-cleaved form (PCSK9_55).
Lijuan Du et al.
Nature communications, 13(1), 3482-3482 (2022-06-18)
How signaling proteins generate a multitude of information to organize tissue patterns is critical to understanding morphogenesis. In Drosophila, FGF produced in wing-disc cells regulates the development of the disc-associated air-sac-primordium (ASP). Here, we show that FGF is Glycosylphosphatidylinositol-anchored to
Fangyuan Chen et al.
Molecular systems biology, 17(8), e10239-e10239 (2021-08-03)
Understanding the mechanism of SARS-CoV-2 infection and identifying potential therapeutics are global imperatives. Using a quantitative systems pharmacology approach, we identified a set of repurposable and investigational drugs as potential therapeutics against COVID-19. These were deduced from the gene expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service