Skip to Content
Merck

344824

α1-3,4-Fucosidase, Xanthomonas sp.

Native α1-3,4-fucosidase from Xanthomonas species. Catalyzes the hydrolysis of α1,3- and α1,4-linked branched, non-reducing terminal fucose from complex carbohydrates. Note: 1 mU = 1 milliunit.

Synonym(s):

α1-3,4-Fucosidase, Xanthomonas sp., α-L-Fucoside Fucohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
EC Number:
NACRES:
NA.54
UNSPSC Code:
12352202
MDL number:
Specific activity:
≥100 mU/mL, ≥400 mU/mg protein
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

specific activity

≥100 mU/mL, ≥400 mU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

N-acetylglucosaminidase, α- and β-galactosidase, neuraminidases, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Native α1-3,4-fucosidase from Xanthomonas species. Catalyzes the hydrolysis of α1,3- and α1,4-linked branched, non-reducing terminal fucose from complex carbohydrates. The presence of sialic acid (but not fucose) linked to galactose will block cleavage.
Note: 1 mU = 1 milliunit.

Physical form

In 25 mM NaCl, 20 mM Tris-HCl, pH 7.5.

Other Notes

One unit is defined as the amount of enzyme that will catalyze the release of 1.0 µmol fucose from 3-fucosyllactose per min at 37°C, pH 5.0.
VanGisbergen, K.P.J.M., et al. 2005. J. Exp. Med.201, 1281.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library