Skip to Content
Merck
All Photos(1)

Key Documents

341006

Sigma-Aldrich

EDAC, Hydrochloride

EDAC HCl is a water-soluble derivative of carbodiimide useful for conjugating haptens to proteins and polypeptides. Used to modify NMDA receptors and as a condensing agent in peptide synthesis.

Synonym(s):

EDAC, Hydrochloride, EDCI, 1-Ethyl-3-(3ʹ-dimethylaminopropyl)carbodiimide, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N3 · xHCl
CAS Number:
Molecular Weight:
155.24 (free base basis)
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

description

RTECS - FF2200000

Assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

aqueous buffer: 2-5 mg/mL
water: 2-5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Cl.N(CCCN(CC)C#N)(C)C

InChI

1S/C8H17N3.ClH/c1-4-11(8-9)7-5-6-10(2)3;/h4-7H2,1-3H3;1H

InChI key

FDXPUDRRFDHONO-UHFFFAOYSA-N

General description

Molar absorptivity (~212 nm; 50 mM phosphate buffer, pH 8):≥6800 M-1cm-1.
Water-soluble carbodiimide derivative. Used to modify NMDA receptors. Also useful for conjugating haptens to proteins or polypeptides.
Water-soluble derivative of carbodiimide useful for conjugating haptens to proteins and polypeptides. Used to modify NMDA receptors and as a condensing agent in peptide synthesis. The major advantage of EDAC coupling is the easy removal of excess reagent and the corresponding urea by washing with dilute acid or water. Carbodiimides catalyze the formation of amide bonds, carboxylic acids, and amines by activating the carboxylate to form an O-acylurea. This intermediate can be attacked by an amine directly to form an amide. EDAC is released as a soluble urea derivative.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Preparation Note

Avoid amine containing buffers.

Reconstitution

Unstable in solution. Reconstitute just prior to use.

Other Notes

Chazot, P.L., et al. 1993. Biochem. Pharmacol. 45, 605.
Richardson, A., et al. 1992. Biochem. Pharmacol. 43, 1415.
Taniuchi, M., et al. 1986. Proc. Natl. Acad. Sci. USA83, 1950.
Chase, J.W., et al. 1983. Proc. Natl. Acad. Sci. USA80, 5480.
Williams, A., et al. 1981. J. Am. Chem. Soc.103, 7090.
Yamada, H., et al. 1981. Biochemistry20, 4836.
Thomas, J.O., et al. 1978. J. Mol. Biol.123, 149.
Ozawa, H. 1970. Biochemistry9, 2158.
Kopple, K.D., et al. 1962. J. Am. Chem. Soc.84, 4457.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Stomach,large intestine,lymph node

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergi Junyent et al.
STAR protocols, 2(3), 100813-100813 (2021-09-28)
These protocols investigate the interaction of cytonemes with localized Wnt. Cell-niche signaling between naive or primed mouse embryonic stem cells (ESCs) and either Wnt-secreting trophoblast stem cells (TSCs) or Wnt signals tethered to microbeads can be scrutinized in vitro. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service