Skip to Content
Merck

324682

Elastase, Porcine Pancreas, High Purity, Crystallized

Elastase, Porcine Pancreas, High Purity, Crystallized, CAS 39445-21-1, is a serine protease that hydrolyzes proteins and peptides, especially at bonds adjacent to neutral amino acid residues.

Synonym(s):

Elastase, Porcine Pancreas, High Purity, Crystallized

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54
MDL number:
Specific activity:
≥25 units/mg protein
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

description

Merck USA index - 14, 3535

form

lyophilized

specific activity

≥25 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

solubility

distilled water: 10 mg/mL

foreign activity

Chymotrypsin ≤0.2%

shipped in

ambient

storage temp.

−20°C

General description

Native elastase from porcine pancreas. A serine protase that catalyzes the hydrolysis of proteins and peptides (especially at bonds adjacent to neutral amino acid residues), including albumin, casein, denatured collagen, elastin, fibrin, and hemoglobin and of a number of synthetic substrates containing aspartic acid, phenylalanine, or tyrosine. Inhibited by DFP, elastinal, and α2-macroglobulin.
Native elastase from porcine pancreas. Catalyzes the hydrolysis of proteins and peptides (especially at bonds adjacent to neutral amino acid residues), including albumin, casein, denatured collagen, elastin, fibrin, and hemoglobin, and of a number of synthetic substrates containing aspartic acid, glutamic acid, phenylalanine, or tyrosine. Preferentially cleaves peptide bonds at the carbonyl end of amino acid residues with small hydrophobic side chains, such as glycine, valine, leucine, isoleucine, and particularly alanine. Inhibited by DFP, elastinal, and α2-macroglobulin. Has an optimal pH of 7.8-8.5; pI = 9.5. Note: Elastase tends to adhere to glass. Hence, use of siliconized glassware is recommended.

Physical form

Lyophilized from 50 mM Trehalose, 1 mM Acetic Acid.

Preparation Note

Following reconstitution, elastase may be stored at 4°C at pH 6.0 for long term use. If incubated at room temperature at or near its optimum pH, elastase rapidly autolyzes to a mixture of peptides. Stock solutions are stable for up to 2 months at 4°C, pH 6.0.

Other Notes

Largman, C. et al. 1976. Biochemestry15, 2491.
Mandl, I. 1962. Methods in Enzymol.5, 665.
One unit is defined as the amount of enzyme that will hydrolyze 1.0 mol of Suc-Ala-Ala-Pro-Abu-pNA (Cat no.324699) per min at 25°C, pH 8.0.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yves Donati et al.
American journal of physiology. Lung cellular and molecular physiology, 318(4), L619-L630 (2020-02-06)
Mouse lung developmental maturation and final alveolarization phase begin at birth. During this dynamic process, alveolar cells modify their morphology and anchorage to the extracellular matrix. In particular, alveolar epithelial cell (AEC) type I undergo cytoplasmic flattening and folding to
Sanja Blaskovic et al.
American journal of physiology. Lung cellular and molecular physiology, 318(4), L606-L618 (2020-01-23)
Harmful consequences of cigarette smoke (CS) exposure during lung development can already manifest in infancy. In particular, early life exposure to nicotine, the main component of CS, was shown to affect lung development in animal models. We aimed to characterize



Global Trade Item Number

SKUGTIN
324682-250U07790788054380
324682-1000U04055977196832