Skip to Content
Merck
All Photos(1)

Documents

324390

Sigma-Aldrich

Anti-Dopamine D₁ Receptor Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

does not contain

preservative

species reactivity

human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DRD1(1812)
rat ... Drd1(24316)

General description

Rabbit polyclonal antibody supplied as lyophilized, undiluted serum. Recognizes the ~50 kDa dopamine D1 receptor protein.
Recognizes the dopamine D1 receptor in rat brain. Does not cross-react with other dopamine receptors.
This Anti-Dopamine D₁ Receptor Antibody is validated for use in ELISA, WB, Frozen Sections, Free-Floating Sections, Paraffin Sections for the detection of Dopamine D₁ Receptor.

Immunogen

Human
a synthetic peptide (MDGTGLVVERDFSC) corresponding to amino acids 9-21 of human D₁dopamine receptor

Application

ELISA (see comments)

Immunoblotting (1:1000)

Frozen Sections (1:5000)

Free-Floating Sections (1:5000)

Paraffin Sections (not recommended)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Reconstitute the antibody with 100 µl PBS containing 1% BSA. For further dilution of the antibody use a similar buffer formulation. Following reconstitution, refrigerate (4°C).

Analysis Note

Positive Control
Rat brain

Other Notes

Mignini, F., et al. 2000. J. Autonom. Pharmacol.20, 325.
This antibody has also been reported to work in ELISA. For blocking immunoblot membranes, use ONLY 4% goat serum or evaporated goat′s milk. For frozen sections a 1:5000 dilution of the reconstituted serum demonstrates specific staining of rat brain sections including the medial septum, nucleus accumbens, dentate gyrus, globus pallidus, medial forebrain bundle, cortex regions 1-3, substantia nigra reticulata, and the ventral tegmental area. Strong nuclear staining of oligodendrocytes was also observed. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye-Min Jeon et al.
Neoplasia (New York, N.Y.), 39, 100894-100894 (2023-03-28)
Recent studies indicate that signaling molecules traditionally associated with central nervous system function play critical roles in cancer. Dopamine receptor signaling is implicated in various cancers including glioblastoma (GBM) and it is a recognized therapeutic target, as evidenced by recent
Florian Giesert et al.
PloS one, 8(5), e63778-e63778 (2013-05-16)
Missense mutations in the leucine-rich repeat kinase 2 gene (LRRK2) are linked to autosomal dominant forms of Parkinson's disease (PD). In order to get insights into the physiological role of Lrrk2, we examined the distribution of Lrrk2 mRNA and different
Dana C Borcherding et al.
PloS one, 6(9), e25537-e25537 (2011-10-04)
Dopamine (DA) binds to five receptors (DAR), classified by their ability to increase (D1R-like) or decrease (D2R-like) cAMP. In humans, most DA circulates as dopamine sulfate (DA-S), which can be de-conjugated to bioactive DA by arylsulfatase A (ARSA). The objective
Jacqueline S Coley et al.
PloS one, 10(2), e0117450-e0117450 (2015-02-04)
Drug abuse is a major comorbidity of HIV infection and cognitive disorders are often more severe in the drug abusing HIV infected population. CD14+CD16+ monocytes, a mature subpopulation of peripheral blood monocytes, are key mediators of HIV neuropathogenesis. Infected CD14+CD16+
Gabriela Assis-de-Lemos et al.
Bioscience reports, 41(12) (2021-11-26)
Dopamine signaling has numerous roles during brain development. In addition, alterations in dopamine signaling may be also involved in the pathophysiology of psychiatric disorders. Neurodevelopment is modulated in multiple steps by reactive oxygen species (ROS), byproducts of oxidative metabolism that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service