Skip to Content
Merck
All Photos(1)

Key Documents

251680

Sigma-Aldrich

Dantrolene, Sodium Salt

A cell-permeable blocker of intracellular calcium release from the sarcoplasmic reticulum that acts by binding to the ryanodine receptor.

Synonym(s):

Dantrolene, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9N4O5 · Na
CAS Number:
Molecular Weight:
336.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

orange

solubility

DMSO: 3 mg/mL
aqueous base: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C14H10N4O5.Na/c19-13-8-17(14(20)16-13)15-7-11-5-6-12(23-11)9-1-3-10(4-2-9)18(21)22;/h1-7H,8H2,(H,16,19,20);/q;+1/p-1/b15-7+;

InChI key

KSRLIXGNPXAZHD-HAZZGOGXSA-M

General description

A cell-permeable blocker of intracellular calcium release from the sarcoplasmic reticulum that acts by binding to the ryanodine receptor. Inhibits long-term depression and depotentiation of synaptic transmission. Has neuroprotective effects in cases of cerebral ischemia.
A skeletal muscle relaxant. Blocks intracellular calcium release from the sarcoplasmic reticulum during excitation-contraction coupling by binding to the ryanodine receptor. Blocks long-term depression and depotentiation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Ryanodine receptor
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wei, H., and Perry, D.C. 1996. J. Neurochem. 67, 2390.
O’Mara, S.M., et al. 1995. Neuroscience68, 621.
Parness, J., and Palnitkar, S.S. 1995. J. Biol. Chem.270, 18465.
Zhang, H., et al. 1995. Am. J. Physiol. 269, H1874.
Hotchkiss, R.S., and Karl, I.E. 1994. Proc. Natl. Acad. Sci. USA91, 3039.
Song, S.K., et al. 1994. Proc. Natl. Acad. Sci. USA91, 3039.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher B Sylvester et al.
Frontiers in cardiovascular medicine, 10, 1112965-1112965 (2023-04-18)
Calcific aortic valve disease (CAVD), a fibrocalcific thickening of the aortic valve leaflets causing obstruction of the left ventricular outflow tract, affects nearly 10 million people worldwide. For those who reach end-stage CAVD, the only treatment is highly invasive valve
Joseph E Pick et al.
The EMBO journal, 36(2), 232-244 (2016-11-20)
mGluR long-term depression (mGluR-LTD) is a form of synaptic plasticity induced at excitatory synapses by metabotropic glutamate receptors (mGluRs). mGluR-LTD reduces synaptic strength and is relevant to learning and memory, autism, and sensitization to cocaine; however, the mechanism is not
Sarayu Ramakrishna et al.
Life science alliance, 7(8) (2024-05-16)
Calcium signaling is integral for neuronal activity and synaptic plasticity. We demonstrate that the calcium response generated by different sources modulates neuronal activity-mediated protein synthesis, another process essential for synaptic plasticity. Stimulation of NMDARs generates a protein synthesis response involving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service