Skip to Content
Merck

233100

CL-387,785

A cell-permeable and irreversible inhibitor of EGF-receptor (EGFR) kinase activity in vivo (IC50 = 250-490 pM) as well as EGF-stimulated autophosphorylation of tyrosine residues in the EGFR in vivo (IC50 = 5 nM).

Synonym(s):

CL-387,785, N-[4-[(3-Bromophenyl)amino]-6-quinazolinyl]-2-butynamide, EKI-785

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H13BrN4O
CAS Number:
Molecular Weight:
381.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

color

pale yellow

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Brc1cc(ccc1)Nc2ncnc3c2cc(cc3)NC(=O)C#CC

InChI

1S/C18H13BrN4O/c1-2-4-17(24)22-14-7-8-16-15(10-14)18(21-11-20-16)23-13-6-3-5-12(19)9-13/h3,5-11H,1H3,(H,22,24)(H,20,21,23)

InChI key

BTYYWOYVBXILOJ-UHFFFAOYSA-N

General description

A cell-permeable and irreversible inhibitor of EGF-receptor (EGFR) kinase activity in vivo (IC50 = 250-490 pM) as well as EGF-stimulated autophosphorylation of tyrosine residues in the EGFR in vivo (IC50 = 5 nM). Blocks EGF-mediated growth in A431 cells. Inhibits proliferation of EGFR or c-ErbB2 expressing cells (IC50 = 31-125 nM) by covalently binding to the EGFR.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR kinase activity
Product does not compete with ATP.
Reversible: no
Target IC50: 250-490 pM inhibiting EGF-receptor (EGFR) kinase activity in vivo; 5 nM inhibiting EGF-stimulated autophosphorylation of tyrosine residues in the EGFR in vivo

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Sweeney, W.E., et al. 2000. Kidney Int.57, 33.
Discafani, C.M., et al. 1999. Biochem. Pharmacol. 57, 917.
Sweeney, W.E., et. al. 1999. Kidney Int.56, 406.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library