Skip to Content
Merck

230790

Chymostatin

Selective neurokinin-1 (NK1) receptor antagonist, solid,powder

Synonym(s):

Chymostatin, [(S)-1-Carboxy-2-phenylethyl]-carbamoyl-α-[2-amidohexahydro-4(S)-pyrimidyl]-(S)-glycyl-[A = Leu; B = Val; or C = Ile]-phenylalaninal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Form:
solid, powder
Quality level:
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chymostatin, Contains chymostatin "A" as a major form.

Quality Level

description

RTECS - GC3047700

form

solid, powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to slightly yellow

solubility

5% glacial acetic acid: 10 mg/mL, DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N7C(CCNC7=N)[C@H](NC(=O)N[C@@H](Cc9ccccc9)C(=O)O)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](Cc8ccccc8)C=O.N4C(CCNC4=N)[C@H](NC(=O)N[C@@H](Cc6ccccc6)C(=O)O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc5ccccc5)C=O.N1C(CCNC1=N)[C@H](NC(=O)N[C@@H](Cc3ccccc3)C(=O)O)C(=O)N[C@@

InChI

1S/2C31H41N7O6.C30H39N7O6/c1-19(2)15-24(27(40)34-22(18-39)16-20-9-5-3-6-10-20)35-28(41)26(23-13-14-33-30(32)36-23)38-31(44)37-25(29(42)43)17-21-11-7-4-8-12-21;1-3-19(2)25(27(40)34-22(18-39)16-20-10-6-4-7-11-20)37-28(41)26(23-14-15-33-30(32)35-23)38-31(44)36-24(29(42)43)17-21-12-8-5-9-13-21;1-18(2)24(26(39)33-21(17-38)15-19-9-5-3-6-10-19)36-27(40)25(22-13-14-32-29(31)34-22)37-30(43)35-23(28(41)42)16-20-11-7-4-8-12-20/h3-12,18-19,22-26H,13-17H2,1-2H3,(H,34,40)(H,35,41)(H,42,43)(H3,32,33,36)(H2,37,38,44);4-13,18-19,22-26H,3,14-17H2,1-2H3,(H,34,40)(H,37,41)(H,42,43)(H3,32,33,35)(H2,36,38,44);3-12,17-18,21-25H,13-16H2,1-2H3,(H,33,39)(H,36,40)(H,41,42)(H3,31,32,34)(H2,35,37,43)/t22-,23?,24-,25-,26-;19-,22-,23?,24-,25-,26-;21-,22?,23-,24-,25-/m000/s1

InChI key

QJIJPLFVFWXMGK-KSPFWWQASA-N

General description

Cell permeable: No

Product does not compete with ATP.

Reversible: No

Chymostatin is a naturally occurring inhibitor of serine proteases with chymotryptic-like specificity. This tetrapeptide inhibitor is produced by various species of Streptomyces bacteria. Chymostatin is used as an inhibitor of MC chymase and is also capable of inhibiting several other proteases, including chymotrypsin, papain, chymotrypsin-like serine proteinases, and lysosomal cysteine proteinases such as cathepsins B, H, andL. Itcontains chymostatin "A" as a major form.

Application

Chymostatin has been used as a:

  • cysteine protease inhibitor in thesubstrate to measure the activated FX (FXa) activity
  • protease inhibitor in enzyme purificationlysis buffer
  • protease inhibitor for ISW2 protein complexpurification

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Chymostatin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Lisa Marquardt et al.
Autophagy, 19(1), 278-295 (2022-05-17)
The yeast PROPPIN Atg18 folds as a β-propeller with two binding sites for phosphatidylinositol-3-phosphate (PtdIns3P) and PtdIns(3,5)P2 at its circumference. Membrane insertion of an amphipathic loop of Atg18 leads to membrane tubulation and fission. Atg18 has known functions at the



Global Trade Item Number

SKUGTIN
230790-5MG04055977218282
230790-25MG04055977218275
230790-10MG04055977218268