Skip to Content
Merck

219372

Cathepsin G Inhibitor I

≥98% (HPLC), solid, cathepsin G inhibitor, Calbiochem®

Synonym(s):

Cathepsin G Inhibitor I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H33N2O6P
CAS Number:
Molecular Weight:
620.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cathepsin G Inhibitor I, The Cathepsin G Inhibitor I, also referenced under CAS 429676-93-7, controls the biological activity of Cathepsin G. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Quality Segment

assay

≥98% (HPLC)

form

solid

potency

53 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 5 mg/mL, methanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)(O)(O)C(c5c6c(ccc5)cccc6)C(=O)c1cc2c(cc1C(=O)N(C3CCN(CC3)C(=O)c4ccccc4)C)cccc2

InChI

1S/C36H33N2O6P/c1-37(28-18-20-38(21-19-28)35(40)25-11-3-2-4-12-25)36(41)32-23-27-14-6-5-13-26(27)22-31(32)33(39)34(45(42,43)44)30-17-9-15-24-10-7-8-16-29(24)30/h2-17,22-23,28,34H,18-21H2,1H3,(H2,42,43,44)

InChI key

GNOZQRKYZJSIPZ-UHFFFAOYSA-N

General description

A potent, selective, reversible, and competitive non-peptide inhibitor of cathepsin G (IC50 = 53 nM and Ki = 63 nM). Weakly inhibits chymotrypsin (Ki = 1.5 µM) and does not have any significant inhibitory effect on thrombin, factor Xa, factor IXa, plasmin, trypsin, tryptase, proteinase 3, and human leukocyte elastase (IC50 >100 µM).
A potent, selective, reversible, competitive, non-peptide inhibitor of cathepsin G [IC50 = 53 nM and Ki = 63 nM]. Weakly inhibits chymotrypsin (Ki = 1.5 µM) and poorly inhibits thrombin, factor Xa, factor IXa, plasmin, trypsin, tryptase, proteinase 3, and human leukocyte elastase (IC50 >100 µM).

Biochem/physiol Actions

Target Ki: 63 nM for cathepsin G
Product does not compete with ATP.
Reversible: yes
Primary Target
cathepsin G
Cell permeable: no

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Greco, M.N., et al. 2002. J. Am. Chem. Soc.124, 3810.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library