Skip to Content
Merck
All Photos(1)

Key Documents

178501

Sigma-Aldrich

Palmostatin B

≥95% (HPLC), solid, APT1 inhibitor, Calbiochem®

Synonym(s):

APT1 Inhibitor, palmostatin B, APT1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36O4
Molecular Weight:
376.53
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

APT1 Inhibitor, palmostatin B, The APT1 Inhibitor, palmostatin B controls the biological activity of APT1.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, beta-lactone acyl protein thioesterase 1 (APT1) inhibitor (IC50 = 0.67 µM, in an enzymatic assay) that is shown to specifically block Ras depalmitoylation, without affecting Ras acylation, in MDCK cells, both in vitro and in vivo. It induces a marked redistribution of NRas to endomembranes (1 µM) without notable cytotoxicity, and is shown to elicit a loss of the precise steady-state localization of palmitoylated Ras proteins in the same cell line. At 50 µM, this inhibitor displays a partial phenotypic reversion in oncogenic HRasG12V-transformed fibroblasts. Furthermore, it demonstrates selectivity for APT1 over phospholipase A1, A2, Cβ and D. It′s inhibitory effect is demonstrated to be consistent with that of APT1 downregulation by siRNA. Also available as a 50 mM solution in DMSO (Cat. No. 508738).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Dekker, F. and Hedberg, C. 2011. Bioorg. Med. Chem. Lett.19, 1376.
Dekker, F., et al. 2010. Nat. Chem. Biol.6, 449.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Coral Y Zhou et al.
eLife, 12 (2023-04-25)
During the rapid and reductive cleavage divisions of early embryogenesis, subcellular structures such as the nucleus and mitotic spindle scale to decreasing cell size. Mitotic chromosomes also decrease in size during development, presumably to scale coordinately with mitotic spindles, but
Yan Sun et al.
Cancer research, 83(12), 2034-2051 (2023-04-20)
Tyrosine kinase inhibitors (TKI) that can suppress the VEGF signaling pathway and angiogenesis have been developed to impede the progression of malignant tumors and have been approved as first-line targeted agents for clear cell renal cell carcinoma (ccRCC). Dysregulation of
Claudia Musial et al.
Redox biology, 55, 102395-102395 (2022-07-17)
Lung cancer is one of the most common cancers worldwide, causing nearly one million deaths each year. Herein, we present the effect of 2-methoxyestradiol (2-ME), the endogenous metabolite of 17β-estradiol (E2), on non-small cell lung cancer (NSCLC) cells. We observed
Xue Huang et al.
Molecular cell, 83(19), 3485-3501 (2023-10-07)
p62 is a well-characterized autophagy receptor that recognizes and sequesters specific cargoes into autophagosomes for degradation. p62 promotes the assembly and removal of ubiquitinated proteins by forming p62-liquid droplets. However, it remains unclear how autophagosomes efficiently sequester p62 droplets. Herein
Zhixing Jin et al.
Reproductive sciences (Thousand Oaks, Calif.) (2023-08-21)
Despite its prevalence and the severity of symptoms, little is known about the pathogenesis and etiology of adenomyosis. In our previous study, Scribble localization has been found to be partially translocated to cytoplasm; however, its regulatory mechanism is known. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service