Skip to Content
Merck

178452

Apolipoprotein A-I, Human Plasma, High-Density Lipoprotein

Native apolipoprotein A-I from human plasma. Functions as a cofactor for lecithin-cholesterol acyltransferase. A component of high density lipoprotein.

Synonym(s):

Apo A-I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12352202
Form:
liquid
Assay:
≥95% (SDS-PAGE)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

Functions as a cofactor of lecithin cholesterol acyltransferase (LCAT).
Native apolipoprotein A-I from human plasma. Functions as a cofactor for lecithin-cholesterol acyltransferase (LCAT). Plays an important role in HDL metabolism.

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 10 mM NH₄HCO₃, pH 7.4.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C). Store at a concentration ≥1 mg/ml.
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Other Notes

Bergot, C., et al. 2001. Eur. J. Biochem.268, 3523.
Pászty, C., et al. 1994. J. Clin. Invest.94, 899.
Breslow, J.L. 1993. Proc. Natl. Acad. Sci. USA90, 8314.
Schultz, J.R., et al. 1992. J. Biol. Chem.267, 21630.
Walsh, A., et al. 1989. J. Biol. Chem.264, 6488.
Brewer, H.B. 1986. Methods Enzymol. 128, 223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library