Skip to Content
Merck
All Photos(2)

Key Documents

16-105

Sigma-Aldrich

Anti-Phosphotyrosine Antibody, clone 4G10®, HRP conjugate

clone 4G10®, Upstate®, from mouse

Synonym(s):

Anti-phosphotyrosine antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.56

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G10®, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

Gene Information

human ... PID1(55022)

General description

Anti-Phosphotyrosine antibody (05-321), cross-linked to horseradish peroxidase
Some of the tyrosine residues can be tagged with a phosphate group (phosphorylated) by protein kinases. (In its phosphorylated state, it is referred to as phosphotyrosine). Tyrosine phosphorylation is considered as one of the key steps in signal transduction and regulation of enzymatic activity. The advent of anti-phospho-tyrosine antibodies is one of significant events in signal transduction research. Before the availability of anti-phosphotyrosine antibodies, tyrosyl phospyhorylation of proteins and enzymes was investigated through hazardous and time-consuming radioactive experiments. Anti-phosphotyrosine antibodies are commonly used in western blots after the targeted proteins have been immunoprecipitated to measure the tyrosyl phosphorylation of the proteins. Anti-phosphotyrosine antibodies are also directly used on cell lysate to examine the overall change of tyrosine phosphorylation level in response to various treatments.

Description
IgG2bκ mouse monoclonal antibody produced in vitro by mouse-mouse hybridoma 4G10. Purified by Protein G-Sepharose chromatography and cross-linked to horseradish peroxidase.

Specificity

Recognizes tyrosine-phosphorylated proteins from all species.

Immunogen

Phosphotyramine-KLH.

Application

Anti-Phosphotyrosine Antibody, clone 4G10, HRP conjugate detects level of Phosphotyrosine & has been published & validated for use in ELISA & WB.
For use in applications in which an HRP conjugate is advantageous, such as WB and IP.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Quality

Routinely evaluated in western blot on RIPA lysates from EGF-treated human A431 carcinoma cells.

Western Blot Analysis:
Tested at 0.5-2 μg/mL, with optimal performance at 2 μg/mL (10 μL per 10 mL blocking buffer). This lot detected tyrosine-phosphorylated proteins in RIPA lysates from EGF-treated human A431 carcinoma cells.

Target description

Dependent upon the molecular weight of the tyrosine phosphorylated protein being detected.

Physical form

200 μg of IgG2bκ-HRP conjugate in 100 μL PBS containing 0.05% Kathon® . Sufficient for 10 western blots, if used at the optimum concentration of 2 μg/mL in 10 mL blocking buffer. Liquid.
Protein G-Sepharose Chromatography

Storage and Stability

Stable for 6 months at 2-8°C from date of receipt. NOTE: DO NOT FREEZE. For maximum recovery of product, centrifuge the original vial prior to removing the cap.
NOTE: For optimal performance, remove HRP precipitate by centrifuging vial prior to use.

Analysis Note

Control
Pervanadate-treated human A431 cell extracts, EGF-treated human A431 cells.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
Kathon is a registered trademark of Rohm and Haas Co.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gajanan Sathe et al.
Clinical proteomics, 13, 13-13 (2016-06-17)
Curcumin, derived from the rhizome Curcuma longa, is a natural anti-cancer agent and has been shown to inhibit proliferation and survival of tumor cells. Although the anti-cancer effects of curcumin are well established, detailed understanding of the signaling pathways altered
Jason E Hudak et al.
Nature chemical biology, 10(1), 69-75 (2013-12-03)
The increase of cell surface sialic acid is a characteristic shared by many tumor types. A correlation between hypersialylation and immunoprotection has been observed, but few hypotheses have provided a mechanistic understanding of this immunosuppressive phenomenon. Here, we show that
Impaired dendritic cell function in aging leads to defective antitumor immunity.
Grolleau-Julius, Annabelle, et al.
Cancer Research, 68, 6341-6349 (2008)
Xiao-Pei Yang et al.
Journal of neuroinflammation, 13(1), 274-274 (2016-10-21)
Streptococcus suis serotype 2 (SS2) is an important zoonotic bacterial pathogen in both humans and animals, which can cause high morbidity and mortality. Meningitis is one of the major clinical manifestations of SS2 infection. However, the specific process of SS2
Atheroprotective effect of oleoylethanolamide (OEA) targeting oxidized LDL.
Fan, A; Wu, X; Wu, H; Li, L; Huang, R; Zhu, Y; Qiu, Y; Fu, J; Ren, J; Zhu, C
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service