Skip to Content
Merck
All Photos(1)

Documents

12-140

Sigma-Aldrich

Src Substrate Peptide

Src Substrate Peptide primarily used in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

Quality Level

mol wt

Mw 1670 Da (1.67 kDa)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... SRC(6714)

General description

Synthetic peptide (KVEKIGEGTYGVVYK) corresponding to amino acids 6-20 of p34cdc2.

Biochem/physiol Actions

Protein Target: Src
Target Sub-Family: TK

Quality

Routinely evaluated in a protein kinase assay as a substrate for Src (p60c-src), (14-117) or Lck (p56lck), active (14-379).

Physical form

Lyophilized powder

Storage and Stability

Stable for 2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Y Chang et al.
Molecular and cellular biology, 18(6), 3245-3256 (1998-06-20)
To isolate and characterize proteins that interact with the unique domain and SH3 and SH2 domains of Src and potentially regulate Src activity, we used the yeast two-hybrid assay to screen a human lung fibroblast cDNA library. We identified RACK1
C K Szekeres et al.
The Journal of biological chemistry, 275(49), 38831-38841 (2000-08-23)
12(S)-Hydroxyeicosatetraenoic acid (12(S)-HETE), a 12-lipoxygenase metabolite of arachidonic acid, has multiple effects on tumor and endothelial cells, including stimulation of invasion and angiogenesis. However, the signaling mechanisms controlling these physiological processes are poorly understood. In a human epidermoid carcinoma cell
Kei Sakamoto et al.
The Journal of biological chemistry, 277(14), 11910-11917 (2002-01-26)
The protein serine/threonine kinase Akt/protein kinase B has been recognized as a critical signaling mediator for multiple cell systems. The function of Akt in skeletal muscle is not well understood, and whether contractile activity stimulates Akt activity has been controversial.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service