Skip to Content
Merck
All Photos(2)

Key Documents

1.16754

Supelco

Water

suitable for UV/Vis spectroscopy, EMSURE®

Synonym(s):

DI water, Deionized water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.21
technique(s):
UV/Vis spectroscopy: suitable
bp:
100 °C (lit.)
vapor pressure:
23 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Water, for analysis EMSURE®

vapor pressure

23 hPa ( 20 °C)

Quality Level

product line

EMSURE®

form

liquid

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.01 mg/L Ammonium (NH4)
≤0.01 mg/L Nitrate (NO3)
≤0.01 mg/L Phosphate (PO4)
≤0.01 mg/L Silicate (SiO2)
≤0.05 mg/L Chloride (Cl)
≤0.05 mg/L Fluoride (F)
≤0.08 mg/L Substances reducing potassium permanganate (O)
≤0.1 mg/L Sulfate (SO4)

evapn. residue

≤1 mg/L

ign. residue

≤1 mg/L

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

cation traces

Ag: ≤0.0004 mg/L
Al: ≤0.0020 mg/L
As: ≤0.0020 mg/L
Au: ≤0.0010 mg/L
B: ≤0.0050 mg/L
Ba: ≤0.0010 mg/L
Be: ≤0.0020 mg/L
Bi: ≤0.0010 mg/L
Ca: ≤0.0050 mg/L
Cd: ≤0.0010 mg/L
Co: ≤0.0010 mg/L
Cr: ≤0.0004 mg/L
Cu: ≤0.0004 mg/L
Fe: ≤0.0010 mg/L
Ga: ≤0.0040 mg/L
Ge: ≤0.0010 mg/L
In: ≤0.0010 mg/L
K: ≤0.0050 mg/L
Li: ≤0.0020 mg/L
Mg: ≤0.0050 mg/L
Mn: ≤0.0004 mg/L
Mo: ≤0.0020 mg/L
Na: ≤0.0100 mg/L
Ni: ≤0.0004 mg/L
Pb: ≤0.0010 mg/L
Pd: ≤0.0080 mg/L
Pt: ≤0.0010 mg/L
Sb: ≤0.0010 mg/L
Sn: ≤0.0010 mg/L
Sr: ≤0.0004 mg/L
Ti: ≤0.0010 mg/L
V: ≤0.0010 mg/L
Zn: ≤0.0040 mg/L
Zr: ≤0.0010 mg/L

SMILES string

O

storage temp.

15-25°C

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Cadmium Standard for Atomic Absorption Spectroscopy: An article in Heliyon (2024) explores the pollution indices and correlation of heavy metal contamination, including cadmium, in groundwater near industrial sites. This emphasizes the role of cadmium AAS standards in monitoring environmental pollution and ensuring compliance with safety regulations (Mohan et al., 2024).

  • Trace Metal Analysis in Food Samples: In a Food Chemistry study (2024), cadmium standards are crucial for the development of novel electrochemical strategies for detecting cadmium in grain and water, using advanced materials like MXene@rGO composite aerogels (Dong et al., 2023).

  • Quality Control in Pharmaceutical Research: The need for precise cadmium calibration standards is underlined in another Food Chemistry publication (2024), which discusses simultaneous electrochemical sensing of heavy metals in food, demonstrating their vital role in ensuring the accuracy and reliability of pharmaceutical and biotechnological analyses (Mirzaei Karazan et al., 2024).

Analysis Note

Spec. conductance at 25 °C (at the time of manufacturing): ≤ 1 µS/cm
Evaporation residue: ≤ 1 mg/l
Residue on ignition: ≤ 1 mg/l
Chloride (Cl): ≤ 0.05 mg/l
Fluoride (F): ≤ 0.05 mg/l
Nitrate (NO₃): ≤ 0.01 mg/l
Phosphate (PO₄): ≤ 0.01 mg/l
Sulfate (SO₄): ≤ 0.1 mg/l
Silicate (SiO₂): ≤ 0.01 mg/l
Ammonium (NH₄): ≤ 0.01 mg/l
Ag (Silver): ≤ 0.0004 mg/l
As (Arsenic): ≤ 0.0020 mg/l
Al (Aluminium): ≤ 0.0020 mg/l
Au (Gold): ≤ 0.0010 mg/l
B (Boron): ≤ 0.0050 mg/l
Ba (Barium): ≤ 0.0010 mg/l
Be (Beryllium): ≤ 0.0020 mg/l
Bi (Bismuth): ≤ 0.0010 mg/l
Ca (Calcium): ≤ 0.0050 mg/l
Cd (Cadmium): ≤ 0.0010 mg/l
Co (Cobalt): ≤ 0.0010 mg/l
Cr (Chromium): ≤ 0.0004 mg/l
Cu (Copper): ≤ 0.0004 mg/l
Fe (Iron): ≤ 0.0010 mg/l
Ga (Gallium): ≤ 0.0040 mg/l
Ge (Germanium): ≤ 0.0010 mg/l
In (Indium): ≤ 0.0010 mg/l
K (Potassium): ≤ 0.0050 mg/l
Li (Lithium): ≤ 0.0020 mg/l
Mg (Magnesium): ≤ 0.0050 mg/l
Mn (Manganese): ≤ 0.0004 mg/l
Mo (Molybdenum): ≤ 0.0020 mg/l
Na (Sodium): ≤ 0.0100 mg/l
Ni (Nickel): ≤ 0.0004 mg/l
Pb (Lead): ≤ 0.0010 mg/l
Pd (Palladium): ≤ 0.0080 mg/l
Pt (Platinum): ≤ 0.0010 mg/l
Sb (Antimony): ≤ 0.0010 mg/l
Sn (Tin): ≤ 0.0010 mg/l
Sr (Strontium): ≤ 0.0004 mg/l
Ti (Titanium): ≤ 0.0010 mg/l
V (Vanadium): ≤ 0.0010 mg/l
Zn (Zinc): ≤ 0.0040 mg/l
Zr (Zirconium): ≤ 0.0010 mg/l
Substances reducing potassium permanganate (O): ≤ 0.08 mg/l

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

Preparation of a standard solution for COD/chloride

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

See All

Protocols

Analytical Method: Iodine in table salt

Hydroxymethylfurfural (HMF) in honey – reflectometric determination after after conversion with a barbituric acid derivative and an aminophenazone derivative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service