Skip to Content
Merck
All Photos(1)

Key Documents

1.14678

Supelco

Formaldehyde Test, photometric

photometric, 0.02-8.00 mg/L (HCHO), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Formaldehyde Test, photometric, 0.02-8.00 mg/L (HCHO), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests

specific analyte(s)

formaldehyde

measuring range

0.02-8.00 mg/L (HCHO)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Chromotropic acid)

storage temp.

15-25°C

General description

Method: photometric 0.02 - 8.00 mg/l HCHO Spectroquant®

This Spectroquant® Formaldehyde Reagent Test allows the accurate quantification of the formaldehyde content in aqueous samples such as disinfectants, preservatives, or process wastewater.

Method applied: In sulfuric solution formaldehyde reacts with chromotropic acid to form a violet dye which is determined photometrically.


The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service