Skip to Content
Merck
All Photos(2)

Key Documents

1.12179

Sigma-Aldrich

Glutardialdehyde solution 25%

for electron microscopy, acc. to P.J. Anderson (purified and filled under nitrogen)

Synonym(s):

Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
NACRES:
NA.21
technique(s):
electron microscopy: suitable
application(s):
diagnostic assay manufacturing
hematology
histology
bp:
100.5 °C/1013 hPa
vapor pressure:
0.27 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

0.27 hPa ( 20 °C)

Quality Level

form

liquid

IVD

for in vitro diagnostic use

concentration

24.7-25.7% (alkalimetric)

technique(s)

electron microscopy: suitable

color

colorless

pH

>3.0 (20 °C in H2O)

bp

100.5 °C/1013 hPa

mp

-6 °C

density

1.06 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutardialdehyde solution 25% - for electron microscopy, according to P.J. Anderson (purified and filled under nitrogen) Glutardialdehyde is a fixing agent for specimens destined for morphological and enzyme histochemical analysis of samples of human origin. Especially sensitive material used for enzyme histochemistry, will be fixed with Glutardialdehyde solution 25% according to P.J. Anderson, (purified and filled into ampoules under nitrogen). Sensitive enzymes e.g. endogenous peroxidase can often only be visualized in this way. Alternatively, the 25% Glutardialdehyde solution is used for the fixation of specimen for semi-thin cuttings and electron microscopy. Both are registered as CE / IVD product for applications in diagnostics. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Assay (alkalimetric): 24.7 - 25.7 %
Identity: conforms
Free acid (as glutaric acid): ≤ 0.2 %
Methanol (HS-GC): ≤ 0.3 %
Optical purity (E 280 nm / E 235 nm): ≥ 3.0

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service